Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

RW1929325

Millipore

RW19 Prefilter 293mm 25PK

Sign Into View Organizational & Contract Pricing

Select a Size

25 EA
Inquire

This item is not able to be purchased as a guest user. For further information, please contact us at +1 866 227 3811or

.


Select a Size

Change View
25 EA
Inquire

About This Item

UNSPSC Code:
40161507
eCl@ss:
32031602

This item is not able to be purchased as a guest user. For further information, please contact us at +1 866 227 3811or

.

material

mixed cellulose esters (MCE)
plain filter
web cellulose
white filter

Quality Level

feature

hydrophilic

filter diam.

293 mm

pore size

90.0 μm pore size

pictograms

Flame

signalword

Danger

hcodes

Hazard Classifications

Flam. Sol. 1

Storage Class

4.1B - Flammable solid hazardous materials

wgk_germany

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service