Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PX1805

Supelco

Isopropyl Acetate

Synonym(s):

Acetic Acid Isopropyl Ester

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352111
assay:
≥99% (GC)
bp:
88-90 °C/1013 hPa
vapor pressure:
61 hPa ( 20 °C)
This product is sold exclusively by Millipore via offline channels. Contact Customer Service

agency

meets USP/NF

Quality Level

vapor pressure

61 hPa ( 20 °C)

assay

≥99% (GC)

form

liquid

bp

88-90 °C/1013 hPa

mp

-73 °C

transition temp

flash point 4 °C

solubility

31 g/L

density

0.871-0.873 ((d 20°C/4°C))

storage temp.

room temp

Application

Meets Reagent Specifications for testing USP/NF monographs

Analysis Note

Assay (GC):99% min
Density (d 20°C/4°C):0.871-0.873
Form:Clear liquid
Identity (IR-spectrum):Conforms

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service