Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PF118

Sigma-Aldrich

MMP-2, Mouse Calvariae

Sign Into View Organizational & Contract Pricing


About This Item

assay

≥90% (SDS-PAGE)

Quality Level

form

liquid

contains

≤0.1% sodium azide as preservative

General description

Native MMP-2 from mouse calvariae. Pro-MMP-2 (progelatinase A) is cleaved in vivo by MT2-MMP to yield a 67 kDa intermediate and the 62 kDa active enzyme.
Native MMP-2 from mouse calvariae. Pro-MMP-2 (progelatinase A) is cleaved in vivo by MT2-MMP to yield a 67 kDa intermediate and the 62 kDa active enzyme. Matrix metalloproteinase-2 (MMP-2) is involved in the degradation of several components of the extracellular matrix under various pathological conditions. In cancer tissues, MMP-2 is secreted by a proenzyme (proMMP-2), which is activated by membrane type 2 matrix metalloproteinase (MT2-MMP) on the cell surface. The 72 kDa pro-MMP-2 (progelatinase A) is cleaved by MT2-MMP to yield a 67 kDa intermediate and the 62 kDa active enzyme. Useful for immunoblotting, substrate cleavage assays, and zymography. Product may contain variable trace amount of active MMP-2.

Unit Definition

One unit is defined as the amount of enzyme that can hydrolyze 1 μmol of the substrate N-(2,4)-DNP-Pro-Gln-Gly-Ile-Ala-Gly-Gln-D-Arg-OH per min at 37°C, pH 7.0.

Physical form

In 200 mM NaCl, 50 mM Tris-HCI, 5 mM CaCl₂, 1 μM ZnCl₂, 0.05% Brij®-35 Detergent, pH 7.0.

Reconstitution

Following initial thaw, aliquot and freeze (-70°C).

Legal Information

Brij is a registered trademark of Croda International PLC

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C G Knight et al.
FEBS letters, 296(3), 263-266 (1992-01-27)
(7-methoxycoumarin-4-yl)Acetyl-Pro-Leu-Gly-Leu-(3-[2,4-dinitrophenyl]-L- 2,3-diaminopropionyl)-Ala-Arg-NH2 (Mca-Pro-Leu-Gly-Leu-Dpa-Ala-Arg-NH2) has been synthesised as a fluorogenic substrate for the matrix metalloproteinases. The highly fluorescent 7-methoxycoumarin group is efficiently quenched by energy transfer to the 2,4-dinitrophenyl group. The punctuated metalloproteinase (PUMP, EC 3.4.24.23) cleaves the substrate at the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service