Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

OP146

Sigma-Aldrich

Anti-MDM2 (Ab-6) Mouse mAb (5B10C)

liquid, clone 5B10C, Calbiochem®

Synonym(s):

Anti-Ubiquitin Protein Ligase, Anti-p53 Binding Protein, Ant-Murine Double Minute Chromosome-2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

5B10C, monoclonal

form

liquid

does not contain

preservative

species reactivity

human

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG2a

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MDM2(4193)

General description

Purified mouse monoclonal antibody. Recognizes the ~90 kDa (apparent MW) MMDM2 protein.
Recognizes the ~90 kDa (apparent MW) MDM2 protein in A549 and MCF-7 cells and breast carcinoma tissue.

Immunogen

Human
recombinant human MDM2

Application


Immunoblotting (2 g/ml)
Immunofluorescence (1 g/ml)
Immunoprecipitation (1 g/reaction, see application references)
Paraffin Sections (2.5 g/ml, heat pre-treatment required)

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Physical form

In 50 mM sodium phosphate buffer, 50% glycerol, pH 7.5.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Analysis Note

Positive Control
A549 cells or breast carcinoma tissue

Other Notes

Antibody should be titrated for optimal results in individual systems.
Marchetti, A., et al. 1995. J. Pathol.175, 31.
Barak, Y., et al. 1993. EMBO. J.12, 461.
Ladanyi, M., et al. 1993. Cancer Res.53, 16.
Leach, F. S., et al. 1993. Cancer Res.53, 2231.
Oliner, J. D., et al. 1993. Nature362, 857.
Momand, J., et al. 1992. Cell69, 1237.
Oliner, J. D., et al. 1992. Nature358, 80.
Fakharzadeh, S. S., et al. 1991. EMBO J.10, 1565.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lijuan Bai et al.
Anticancer research, 40(11), 6051-6062 (2020-10-29)
Chemoresistance is a major obstacle in the treatment of prostate cancer (PCa). It is imperative to develop novel strategies for overcoming chemoresistance and improving clinical outcomes. We evaluated the in vitro activity and mechanism of action of dihydroergocristine (DHECS), an
Johanna Huun et al.
Translational oncology, 10(5), 806-817 (2017-08-28)
MDM2 is a negative regulator of p53 and is upregulated in numerous human cancers. While different MDM2 splice variants have been observed in both normal tissues and malignant cells, their functions are poorly understood. We evaluated the effect of MDM2
Raymond L Warters et al.
Radiation research, 172(1), 82-95 (2009-07-08)
Although skin is usually exposed during human exposures to ionizing radiation, there have been no thorough examinations of the transcriptional response of skin fibroblasts and keratinocytes to radiation. The transcriptional response of quiescent primary fibroblasts and keratinocytes exposed to from
Kai Yang et al.
Nature communications, 7, 13599-13599 (2016-11-26)
The nucleolus has been recently described as a stress sensor. The nucleoplasmic translocation of nucleolar protein nucleophosmin (NPM1) is a hallmark of nucleolar stress; however, the causes of this translocation and its connection to p53 activation are unclear. Using single

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service