Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

OP104L

Sigma-Aldrich

Anti-p53 (Ab-11) (Pantropic) Mouse mAb (PAb1802)

lyophilized, clone PAb1802, Calbiochem®

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$385.00

$385.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
$385.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$385.00


Check Cart for Availability

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

PAb1802, monoclonal

form

lyophilized

does not contain

preservative

species reactivity

human, mouse

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

isotype

IgG1

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... TP53(7157)

General description

Purified mouse monoclonal antibody generated by immunizing mice with the specified immunogen and fusing splenocytes with NS1 mouse myeloma cells. Recognizes the ~53 kDa wild-type and mutant forms of p53.
Recognizes the ~53 kDa wild-type and mutant p53 protein in immunoblotting. Recognizes only the wild-type p53 protein in immunoprecipitation.
This Anti-p53 (Ab-11) (Pantropic) Mouse mAb (PAb1802) is validated for use in Immunoblotting, IP, Frozen Sections, IF Paraffin Sections for the detection of p53 (Ab-11) (Pantropic).

Immunogen

Epitope: within amino acids 306-393 of human p53
Human
recombinant, human p53 protein

Application

Immunoblotting (2.5 µg/ml, chemiluminescence, see application references)

Immunoprecipitation (1 µg/reaction, see comments, see application references)

Frozen Sections (not recommended)

Immunofluorescence (not recommended)

Paraffin Sections (not recommended)

Warning

Toxicity: Standard Handling (A)

Physical form

Lyophilized from a volatile buffer, 100 µg BSA.

Reconstitution

Reconstitute the lyophilized antibody with sterile PBS, pH 7.4, or sterile 20 mM Tris-saline (20 mM Tris containing 0.15 M NaCl), pH 7.4, to yield a final concentration of 100 µg/ml; product will be more stable if 0.1% sodium azide is included. Lyophilized antibodies should be reconstituted at 4°C by gradual addition of solvent over a period of 1-2 h with occasional gentle mixing.

Analysis Note

Negative Control
SK-OV-3 cells
Positive Control
A431 or SK-BR-3 cells

Other Notes

Levine, A.J. 1997. Cell88, 323.
Miller, S.D., et al. 1997. Mol. Cell. Biol.17, 2194.
Pellegata, N.S., et al. 1996. Proc. Natl. Acad. Sci. USA93, 15209.
Sturzbecher, H.-W., et al. 1996. EMBO J.15, 1992.
Argarwal, M.L., et al. 1995. Proc. Natl. Acad. Sci. USA92, 8493.
Götz, C. and Montenarh, M. 1995. Int. J. Oncology6, 1129.
Reed, M. et al. 1995. Proc. Natl. Acad. Sci. USA92, 9455.
Waldman, T., et al. 1995. Cancer Res.55, 5187.
Clarke, A.R., et al. 1994. Oncogene9, 1767.
Cho, Y., et al. 1994. Science265, 346.
El-Deiry, W.S., et al. 1994. Cancer Res.54, 1169.
Greenblatt, M.S., et al. 1994. Cancer Res.54, 4855.
Lane, D.P. 1992. Nature358, 15.
This antibody will immunoprecipitate wild-type p53 only. This antibody will stain methanol-fixed cells. Antibody should be titrated for optimal results in individual systems.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kimberley J Botting et al.
Journal of the American Heart Association, 3(4) (2014-08-03)
Placental insufficiency is the leading cause of intrauterine growth restriction in the developed world and results in chronic hypoxemia in the fetus. Oxygen is essential for fetal heart development, but a hypoxemic environment in utero can permanently alter development of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service