Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MXSMCYM48

Millipore

Milliflex Cassette prefilled with Yeast and Mold Agar

This medium is recommended for the isolation and cultivation of yeasts, molds and other aciduric microorganisms

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116129
eCl@ss:
32160801
NACRES:
NA.74

form

plate (pre-poured agar)

shelf life

6 mo.

feature

ready-to-use

packaging

cassette of 10 mL
pack of 48 tests

manufacturer/tradename

Milliflex®

color

light ambermedium, very slightly opalescent

input

sample type beer
sample type bottled water
sample type pharmaceutical(s)
sample type cosmetics

pH

6.2±0.2 (25.0 °C)

application(s)

bioburden testing
cosmetics
microbiology
pharmaceutical
water monitoring

compatibility

for use with Milliflex®

shipped in

ambient

storage temp.

2-8°C

suitability

molds
yeasts

General description

By combining the Milliflex products, accessories and services you can immediately streamline your microbial contamination testing workflow, leverage your quality assurance level by avoiding risks of exogenous contamination and ensure compliance to all GMP/GLP requirements.

Application

Type of Test
Bioburden

Microbial Limits

Packaging

10 mL Agar in plastic Cassette

Specifications

Incubation Temperature: 20–30 °C
Incubation Time: 18–72 hrs

Analysis Note

Colonies typically appear white to beige
QC organisms
C. albicans (ATCC 10231)

S. cerevisiae (ATCC 7754)
Target Microorganisms: Yeast and Mold

Legal Information

MILLIFLEX is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service