sample type pharmaceutical(s) sample type bottled water sample type cosmetics sample type beer
application(s)
bioburden testing water monitoring
compatibility
Milliflex® Plus Pump
shipped in
ambient
General description
The Milliflex® Plus Printer is connected to the Milliflex® Plus vacuum pump, allowing all relevant data to be tracked and printed for easy report generation after filtration.
Application
The Milliflex® Plus Printer is an accessory of the Milliflex® Plus vacuum pump used during filtration of the test samples for general microbiological analysis, bioburden testing, and water monitoring.
Linkage
Replaces: MXPRINT01
Legal Information
MILLIFLEX is a registered trademark of Merck KGaA, Darmstadt, Germany
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.