Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

MXGASKTMM

Gasket kit

for use with Milliflex® PLUS, for use with Milliflex Oasis® funnel

Sign Into View Organizational & Contract Pricing

Select a Size

MXGASKTMM
$114.00

$114.00


Check Cart for Availability

Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
MXGASKTMM
$114.00

About This Item

UNSPSC Code:
41116129

$114.00


Check Cart for Availability

Notify Me

Get notified when this item is ready to ship via email.

material

silicon

packaging

pkg of 5 units (per pack)

manufacturer/tradename

Milliflex Oasis®

application(s)

bioburden testing
food and beverages
pharmaceutical
water monitoring

compatibility

for use with Milliflex Oasis® funnel
for use with Milliflex® PLUS

shipped in

ambient

General description

Gasket kit for Milliflex® Plus head for Milliflex Oasis® funnel is an additional accessory and allows the use of Milliflex Oasis® funnel with the Milliflex® Plus head. The Milliflex Oasis® filtration unit is used for bioburden testing and has been designed to ensure optimal microbial recovery. The sterilized, disposable filtration devices streamline the workflow, save time and gives highly reliable microbial enumeration results. The Milliflex® Plus head enables easy attachment of the Milliflex® Plus pump to the Milliflex Oasis® funnel. The spare part gasket kit contains spare parts for five replacements and allows to maintain the filtration head when requested.

Application

Gasket kit for Milliflex® Plus head for Milliflex Oasis® funnel is an accessory to be used in Milliflex Oasis® filtration pump and finds its application in bioburden testing and water monitoring of beer, bottled water, cosmetics and Pharmaceutical products analysis.

Legal Information

MILLIFLEX is a registered trademark of Merck KGaA, Darmstadt, Germany
MILLIFLEX OASIS is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service