Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MX1583G

Supelco

Molecular Sieve, Type 4A

bead diam. 1/16 in.

Synonym(s):

Molecular Sieve

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23201100

form

pellets

bead diam.

1/16 in.

shipped in

ambient

storage temp.

room temp

General description

1/16 Beads

Application

for Drying

Analysis Note

Color: Tan
Diameter: 1/16 inchinch
Form: Pellets
Identification (sodium): To pass test

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class

8B - Non-combustible corrosive hazardous materials

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service