Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MVHABG124

Millipore

Microfil V, 100mL, 0.45µm Mixed Cellulose Esters black gridded

Drinking water

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116129
NACRES:
NB.24

material

black filter
gridded filter
mixed cellulose esters (MCE)
polypropylene

sterility

sterile; ethylene oxide treated

manufacturer/tradename

Microfil®

parameter

100 mL sample volume

capacity

100 mL

filter diam.

47 mm

funnel capacity

100 mL

matrix

MF-Millipore

pore size

0.45 μm pore size

input

sample type beverage(s)
sample(s) (environmental)
sample type food(s)
sample type soft drink
sample type beer
sample type bottled water
sample type sport drink
sample type wine
sample type cider
sample type cosmetics

application(s)

bioburden testing
clinical
cosmetics
environmental
environmental
food and beverages
industrial qc
pharmaceutical

shipped in

ambient

Application

Drinking water

Legal Information

MF-Millipore is a trademark of Merck KGaA, Darmstadt, Germany
MICROFIL is a registered trademark of Merck KGaA, Darmstadt, Germany

pictograms

Flame

signalword

Danger

hcodes

Hazard Classifications

Flam. Sol. 1

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service