Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MR-020P

Sigma-Aldrich

EmbryoMax® KSOM Mouse Embryo Media

(1X), Powder, w/o Phenol Red, 5 x 10ml

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
eCl@ss:
32160801
NACRES:
NA.75

form

powder

manufacturer/tradename

Specialty Media
EmbryoMax®

technique(s)

cell culture | embryo: suitable
cell culture | stem cell: suitable

input

sample type: mouse embryo(s)

General description

We now offer our most popular embryo culture media in a dry powder format. Each has been formulated for optimal, consistent performance. This new dry powdered format offers greatly increased shelf-life, easy and quick preparation, and the freshest nutrients to support the development of your embryos in culture. Just add the sterile diluent (included in the kit) to the powder, mix gently, and filter to prepare FRESH medium with the correct pH and osmolarity.

Packaging

5 x 10 mL

Storage and Stability

Store at 2-8°C for upto 1 year

Legal Information

EmbryoMax is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

pictograms

Exclamation mark

signalword

Warning

Storage Class

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Role of Tet proteins in 5mC to 5hmC conversion, ES-cell self-renewal and inner cell mass specification.
Ito S. et al.
Nature null

Articles

Advanced KSOM mouse embryo media that can be used as a single medium for both harvest and culture of mouse embryos to facilitate creation of transgenic knockout mice.

Mouse embryo media and embryo validated reagents for transgenic mouse embryo culture

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service