Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MMQTV0KT4

Milliflex Oasis® Quantum Kit

Including Milliflex Oasis® HABG funnels, parameter 100 mL, pore size 0.45 μm, (Black gridded membrane)

Synonym(s):

Fluorescence-based rapid detection system

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
Pricing and availability is not currently available.

material

mixed cellulose esters (MCE)

parameter

100 mL

volume

47 mm

pore size

0.45 μm

General description

Milliflex® Quantum Universal Kit for Milliflex Oasis® is a fluorescence-based technology for the quantitative and rapid detection of microorganisms in filterable samples. The universal enzymatic fluorescent staining is non-destructive, allowing the identification of the contaminant following a positive result. Therefore, the Milliflex® Quantum system is a fast and reliable alternative for the detection of microorganisms. The kit includes fluorescent reagents and Milliflex Oasis® HABG 100 mL funnels (0.45 μm membrane, Black gridded).

Application

The Milliflex® Quantum Universal Kit for Milliflex Oasis® is used for rapid bioburden testing in filtrable samples, including in-process and final product samples (pharmaceuticals and water). The product is also used in industrial QC and water monitoring.

Legal Information

MILLIFLEX is a registered trademark of Merck KGaA, Darmstadt, Germany
MILLIFLEX OASIS is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Milliflex® Quantum system enables rapid microbial contamination detection with fluorescence staining technology.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service