MMQTV0KT3
Milliflex Oasis® Quantum Kit
Including Milliflex Oasis® GSWG funnels, parameter 100 mL, pore size 0.22 μm, (White gridded membrane)
Synonym(s):
Fluorescence-based rapid detection system
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352200
material
mixed cellulose esters (MCE)
parameter
100 mL
volume
47 mm
pore size
0.22 μm
General description
Milliflex® Quantum Universal Kit for Milliflex Oasis® is a fluorescence-based technology for the quantitative and rapid detection of microorganisms in filterable samples. The universal enzymatic fluorescent staining is non-destructive, allowing the identification of the contaminant following a positive result. Therefore, the Milliflex® Quantum system is a fast and reliable alternative for the detection of microorganisms. The kit includes fluorescent reagents and Milliflex Oasis® GSWG 100 mL funnels (0.22 μm membrane, White gridded).
Application
The Milliflex® Quantum Universal Kit for Milliflex Oasis® is used for rapid bioburden testing in filtrable samples, including in-process and final product samples (pharmaceuticals and water). The product is also used in industrial QC and water monitoring.
Legal Information
MILLIFLEX is a registered trademark of Merck KGaA, Darmstadt, Germany
MILLIFLEX OASIS is a registered trademark of Merck KGaA, Darmstadt, Germany
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Milliflex® Quantum system enables rapid microbial contamination detection with fluorescence staining technology.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service