Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

MMHMAG-44K

Millipore

MILLIPLEX® Mouse Metabolic Hormone Magnetic Bead Panel - Metabolism Multiplex Assay

MMHE-44K has replaced MMHMAG-44K. MMHE-44K can be ordered using the "DESIGN AND PRICE YOUR KIT" link below., The analytes available for MMHE-44K are Amylin Active, C-Peptide 2, Ghrelin, GIP, GLP-1 Active or GLP-1 Total, Glucagon, IL-6, Insulin, Leptin, MCP-1, PP, PYY, Resistin, Secretin, and TNFa.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000

feature

MMHE-44K has replaced MMHMAG-44K. MMHE-44K can be ordered using the "DESIGN AND PRICE YOUR KIT" link below.
The analytes available for MMHE-44K are Amylin Active, C-Peptide 2, Ghrelin, GIP, GLP-1 Active or GLP-1 Total, Glucagon, IL-6, Insulin, Leptin, MCP-1, PP, PYY, Resistin, Secretin, and TNFa.

species reactivity

mouse

manufacturer/tradename

Milliplex®

assay range

accuracy: 104%
(Amylin (Active))

accuracy: 84%
(PYY (Total))

accuracy: 88%
(TNFα)

accuracy: 89%
(C-Peptide 2)

accuracy: 92%
(Ghrelin (Active))

accuracy: 96%
(Resistin)

sensitivity: 1-43 pg/mL
standard curve range: 14-10,000 pg/mL
(Glucagon)

standard curve range: 2.7-2,000 pg/mL
(GIP (Total))

standard curve range: 27-10,000 pg/mL
(IL-6 & TNFα)

standard curve range: 41-30,000 pg/mL
(Amylin (Active) & GLP-1 (Active))

standard curve range: 69-50,000 pg/mL
(C-Peptide 2, Insulin, Leptin, MCP-1, & Resistin)

standard curve range: 7-5,000 pg/mL
(Ghrelin (Active), PP & PYY (Total))

technique(s)

multiplexing: suitable

detection method

fluorometric (Luminex xMAP)

shipped in

wet ice

General description

This product has been replaced by our new MMHE-44K. The MILLIPLEX® Mouse Metabolic Hormone Expansion Panel is a 15-plex kit to be used for the simultaneous quantification of any or all of the following analytes in tissue/lysate and culture supernatant samples and mouse serum or plasma: Amylin Active, C-Peptide 2, Ghrelin, GIP, GLP-1 Active or GLP-1 Total, Glucagon, IL-6, Insulin, Leptin, MCP-1, PP, PYY, Resistin, Secretin, and TNFα. This kit uses a 96-well format, contains a lyophilized standard cocktail, two internal assay quality controls and can measure up to 38 samples in duplicate.

The Luminex® xMAP® platform uses a magnetic bead immunoassay format for ideal speed and sensitivity to quantitate multiple analytes simultaneously, dramatically improving productivity while conserving valuable sample volume.

Panel Type: Metabolism

Application

  • This panel has been replaced by MMHE-44K

Features and Benefits

Design your multiplex kit by choosing available analytes within this panel.

Linkage

Replaces: MENDO-75K

Legal Information

Luminex is a registered trademark of Luminex Corp
MILLIPLEX is a registered trademark of Merck KGaA, Darmstadt, Germany
xMAP is a registered trademark of Luminex Corp

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sara C Campbell et al.
PloS one, 11(3), e0150502-e0150502 (2016-03-10)
The gut microbiota is now known to play an important role contributing to inflammatory-based chronic diseases. This study examined intestinal integrity/inflammation and the gut microbial communities in sedentary and exercising mice presented with a normal or high-fat diet. Thirty-six, 6-week
The inflammatory profile and liver damage of a sucrose-rich diet in mice.
Oliveira, LS; Santos, DA; Barbosa-da-Silva, S; Mandarim-de-Lacerda, CA; Aguila, MB
The Journal of Nutritional Biochemistry null
Short-chain fructo-oligosaccharides modulate intestinal microbiota and metabolic parameters of humanized gnotobiotic diet induced obesity mice.
Respondek, F; Gerard, P; Bossis, M; Boschat, L; Bruneau, A; Rabot, S; Wagner, A; Martin, JC
Testing null
Harshita Chodavarapu et al.
PloS one, 8(4), e62833-e62833 (2013-05-07)
Alterations within the renal renin angiotensin system play a pivotal role in the development and progression of cardiovascular and renal disease. Angiotensin converting enzyme 2 (ACE2) is highly expressed in renal tubules and has been shown to be renoprotective in
Pharmacological inhibition of diacylglycerol acyltransferase 1 reduces body weight and modulates gut peptide release--potential insight into mechanism of action.
Liu, J; Gorski, JN; Gold, SJ; Chen, D; Chen, S; Forrest, G; Itoh, Y; Marsh, DJ; McLaren et al.
Obesity (Silver Spring, Md.) null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service