MDE6013FS1
Millistak+® Pod Depth Filter
DE60, filtration area 1.4 m², Process Scale
Synonym(s):
Millistak+ Pod Depth Filter, DE60 media series, 1.4 m 2 surface area, 1/pk
Select a Size
$898.00
Select a Size
About This Item
$898.00
Recommended Products
Product Name
Millistak+® Pod Depth Filter, DE60, cellulose, filtration area 1.4 m2, hydrophilic
material
cellulose
filter aid (inorganic)
glass filled polypropylene
Quality Level
description
For clarification of serum, plasma, vaccines, cell culture or other fluids.
feature
hydrophilic
manufacturer/tradename
Millistak+®
parameter
1.0 bar max. differential pressure (15 psid) at 80 °C (Forward)
1.0 bar max. inlet pressure (15 psid) at 80 °C
2.1 bar max. differential pressure (30 psid) at 25 °C (Forward)
2.1 bar max. differential pressure (30 psid) at 25 °C (Reverse)
3.5 bar max. inlet pressure (50 psid) at 25 °C
H
32 cm (12.5 in.)
L
62 cm (24.2 in.)
filtration area
1.4 m2
impurities
<0.25 EU/mL bacterial endotoxins (LAL test, Aqueous extraction)
input
sample type liquid
conductivity
1.52-1.94 μS/cm at 25 °C (product water)
filter grade
DE60
General description
This DE60 media series delivers optimal performance through a gradient density matri, ideal for primary or secondary clarification applications. Millistak+® Pod filters are available in three distinct series of media grades in order to meet your specific needs - DE, CE and HC.
The Millistak+® Pod system is ideal for a wide variety of primary and secondary clarification applications, including cell cultures, yeast and E. coli lysates post centrifuge, E. coli refolds, media, vaccines, plasma proteins and sera.
Easy to Use
With the compact, modular design of our Millistak+® new Pod system, you can increase productivity and shorten cycle times.
Installation and setup of the Pod system is simple and straightforward. The unique design of the disposable adapters and disposable manifolds makes it easy to connect the Pod filters to rest of the unit operations in the process. The self-contained and disposable nature of the system protects operators from exposure to biohazards and eliminates maintenance as well as cleaning validation requirements.
Application
Features and Benefits
- Low hold-up volume for greater product yield
- Broad range of media types offered in single and multilayer products
- Flexible, modular format offers scalability from 5 to 12,000 liters or more
- Patented disposable design eliminates need for housing, CIP or cleaning validation
- Self-contained Pod filters protect operators from exposure to biohazards
- Robust construction is easy to use and set up
- Smaller footprint facilitates use in tight spaces
Specifications
Sodium: ≤11.5 mg/ft2
Calcium: ≤0.050 mg/ft2
Magnesium: ≤0.129 mg/ft2
Mercury: ≤0.0004 mg/ft2
Chromium: ≤0.001 mg/ft2
Aluminum: ≤0.025 mg/ft2
Potassium: ≤0.351 mg/ft2
Lead: ≤0.0004 mg/ft2
Arsenic: ≤0.003 mg/ft2
Cobalt: ≤0.0002 mg/ft2
Iron: ≤0.042 mg/ft2
Manganese: ≤0.001 mg/ft2
Titanium: <0.001 mg/ft2
Zinc: ≤0.002 mg/ft2
Nickel: ≤0.001 mg/ft2
Copper: ≤0.003 mg/ft2
Preparation Note
1 autoclave cycle of 60 minutes at 123 °C
Analysis Note
Legal Information
Storage Class
10-13 - German Storage Class 10 to 13
wgk_germany
nwg
flash_point_f
Not applicable
flash_point_c
Not applicable
What is the Emprove® Program?
The Emprove® Program is a system providing comprehensive and thorough documentation of our filters and single-use components, pharma raw materials, and starting materials. Four document types are included with an Emprove® Program subscription:
Free of charge - Sign in to download
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service