Skip to Content
MilliporeSigma

MDE5007FS1

Millipore

Millistak+® Pod Depth Filter

DE50, filtration area 0.77 m², Process Scale

Synonym(s):

Millistak+ Pod Depth Filter, DE50 media series, 0.77 m 2 surface area, 1/pk

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$629.00

$629.00


Check Cart for Availability

Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
1 EA
$629.00

About This Item

UNSPSC Code:
23151806
eCl@ss:
32031690

$629.00


Check Cart for Availability

Notify Me

Get notified when this item is ready to ship via email.

Product Name

Millistak+® Pod Depth Filter, DE50, cellulose, filtration area 0.77 m2, hydrophilic

material

cellulose
filter aid (inorganic)
glass filled polypropylene

Quality Level

description

For clarification of serum, plasma, vaccines, cell culture or other fluids.

feature

hydrophilic

manufacturer/tradename

Millistak+®

parameter

1.0 bar max. differential pressure (15 psid) at 80 °C (Forward)
1.0 bar max. inlet pressure (15 psid) at 80 °C
2.1 bar max. differential pressure (30 psid) at 25 °C (Forward)
2.1 bar max. differential pressure (30 psid) at 25 °C (Reverse)
3.5 bar max. inlet pressure (50 psid) at 25 °C

H

32 cm (12.5 in.)

L

62 cm (24.2 in.)

filtration area

0.77 m2

impurities

<0.25 EU/mL bacterial endotoxins (LAL test, Aqueous extraction)

input

sample type liquid

conductivity

1.52-1.94 μS/cm at 25 °C (product water)

filter grade

DE50

General description

The Millistak+® depth filter media is offered in a scalable, disposable format providing greater flexibility because of its unique modular design. The Millistak+® MDE5007FS1 comes in a POD format featuring a 32 cm (H) x 62 cm (L) and a filtration area of 0.77 m2.

This DE50 media series delivers optimal performance through a gradient density matri, ideal for primary or secondary clarification applications. Millistak+® Pod filters are available in three distinct series of media grades in order to meet your specific needs - DE, CE and HC.

The Millistak+® Pod system is ideal for a wide variety of primary and secondary clarification applications, including cell cultures, yeast and E. coli lysates post centrifuge, E. coli refolds, media, vaccines, plasma proteins and sera.

Easy to Use

With the compact, modular design of our Millistak+® new Pod system, you can increase productivity and shorten cycle times.

Installation and setup of the Pod system is simple and straightforward. The unique design of the disposable adapters and disposable manifolds makes it easy to connect the Pod filters to rest of the unit operations in the process. The self-contained and disposable nature of the system protects operators from exposure to biohazards and eliminates maintenance as well as cleaning validation requirements.

Application

For clarification of serum, plasma, vaccines, cell culture or other fluids.

Features and Benefits

  • Low hold-up volume for greater product yield
  • Broad range of media types offered in single and multilayer products
  • Flexible, modular format offers scalability from 5 to 12,000 liters or more
  • Patented disposable design eliminates need for housing, CIP or cleaning validation
  • Self-contained Pod filters protect operators from exposure to biohazards
  • Robust construction is easy to use and set up
  • Smaller footprint facilitates use in tight spaces

Specifications

Filter Grade: DE50
Per 24-hour static soak in 10 L pure water post autoclave (1 cycle of 30 minutes at 123 °C) and pure water flush of 10 L per ft2 (100 L/m2) of surface area.
Sodium: ≤11.5 mg/ft2
Calcium: ≤0.050 mg/ft2
Magnesium: ≤0.129 mg/ft2
Mercury: ≤0.0004 mg/ft2
Chromium: ≤0.001 mg/ft2
Aluminum: ≤0.025 mg/ft2
Potassium: ≤0.351 mg/ft2
Lead: ≤0.0004 mg/ft2
Arsenic: ≤0.003 mg/ft2
Cobalt: ≤0.0002 mg/ft2
Iron: ≤0.042 mg/ft2
Manganese: ≤0.001 mg/ft2
Titanium: <0.001 mg/ft2
Zinc: ≤0.002 mg/ft2
Nickel: ≤0.001 mg/ft2
Copper: ≤0.003 mg/ft2

Preparation Note

Sterilization Method
1 autoclave cycle of 60 minutes at 123 °C

Analysis Note

Product Water Conductivity at 25°C: post autoclave (1 cycle of 60 minutes at 123 °C) and pure water flush of 5 liters per ft2 (50 L/m2) of media surface area

Legal Information

MILLISTAK+ is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

10-13 - German Storage Class 10 to 13

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


What is the Emprove® Program?

The Emprove® Program is a system providing comprehensive and thorough documentation of our filters and single-use components, pharma raw materials, and starting materials. Four document types are included with an Emprove® Program subscription:

Material Qualification Dossier

Free of charge - Sign in to download

Learn more about the benefits an Emprove® Program subscription can provide
Why are these documents unavailable?
Emprove® Dossiers contain information intended to support the registration process and risk assessment activities of pharmaceutical producers and contain confidential information. Please contact us if you would like access.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service