Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MAISA4VP1

Millipore

MAS-100 Iso NT® Validation Protocol (A4)

The MAS-100 Iso NT Sampler validation protocol is available to make validation faster and easier and ensures that your system′s performance remains compliant with its validated specifications.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116500
eCl@ss:
25169090
NACRES:
JA.11
This product is sold exclusively by Millipore via offline channels. Contact Customer Service

Quality Level

manufacturer/tradename

MAS-100®

input

sample type pharmaceutical(s)

application(s)

industrial qc

shipped in

ambient

General description

Service Type: Validation Services

Application

The MAS-100 Iso NT Sampler validation protocol is available to make validation faster and easier and ensures that your system′s performance remains compliant with its validated specifications.

Legal Information

MAS-100 is a registered trademark of MBV AG
MAS-100 ISO NT is a registered trademark of MBV AG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service