Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABX7635

Anti-human C-reactive protein (high sensitivity CRP), clone C135

clone C135, from mouse

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
Pricing and availability is not currently available.

biological source

mouse

antibody form

purified antibody

antibody product type

primary antibodies

clone

C135, monoclonal

species reactivity

human

technique(s)

ELISA: suitable
western blot: suitable

isotype

IgG2b

Gene Information

human ... CRP(1401)

General description

Antibody recognises C-Reactive Protein (CRP) which has long been established as inflammation marker in blood. More recently sophisticated and sensitive tests been developed to make it possilbe to use CRP as a predictive marker for Cardiovascular Disease (CVD) and Coronary Heart Disease (CHD)

application

Confirmed matched pairing. Capture Antibody MABX7630-1KC or MABX7635-1KC; Detector Antibody MABX7635-1KC
Research Category
Cardiac
Western Blot, ELISA

Physical form

Format: Purified
PBS (pH 7.4) with 0.09% sodium azide.
Protein A purified

Storage and Stability

Stable for 12 months at 2-8ºC from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service