Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABX724

Anti-Cystatin C, clone 24

clone 24, from mouse

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
Pricing and availability is not currently available.

biological source

mouse

antibody form

purified antibody

antibody product type

primary antibodies

clone

24, monoclonal

species reactivity

human

technique(s)

ELISA: suitable
western blot: suitable

isotype

IgG1

Gene Information

human ... CST3(1471)

General description

Antibody binds Cystatin C a marker traditionally used in the detection of Chronic Kidney Disease (CKD). More recently there has been increasing evidence that the level of Cystatin C can be linked to hardening of arteries and the deteriorating cardiovascular condition.

Application

Confirmed matched pairing. Capture Antibody MABX724-1KC; Detector Antibody MABX719-1KC or MABX728-1KC
Research Category
Cardiac
Western Blot, ELISA

Physical form

Format: Purified
PBS (pH 7.4) with 0.09% sodium azide.
Protein A purified

Storage and Stability

Stable for 12 months at 2-8ºC from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service