Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABX4152

Anti-human PAPP-A (dPAPP-A) clone 52

clone 52, from mouse

Synonym(s):

dPAPP-A, Pregnancy associated plasma protein A

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
Pricing and availability is not currently available.

biological source

mouse

antibody form

purified antibody

antibody product type

primary antibodies

clone

52, monoclonal

species reactivity

human

technique(s)

ELISA: suitable
western blot: suitable

isotype

IgG1

Gene Information

human ... PAPPA(5069)

General description

Antibody recognises dimeric Pregnancy Associated Plasma Protein A (dPAPP-A). Elevated levels of this protein have been linked with unstable arterosceleric plaques which as made it an interesting candidate for the diagnosis and prognosis of Acute Coronary Syndrome (ACS)
ELISA;Western Blotting

Physical form

Format: Purified

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service