Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABT912

Sigma-Aldrich

Anti-Integrin B8 Antibody, clone11D4

clone 11D4, from mouse

Synonym(s):

ITGB8

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

antibody form

purified antibody

antibody product type

primary antibodies

clone

11D4, monoclonal

species reactivity

human

technique(s)

western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... ITGB8(3696)

General description

Integrin beta-8 (UniProt: P26012) is encoded by the ITGB8 gene (Gene ID: 3696) in human. Integrin complexes mediate cell-cell and cell-extracellular matrix interactions and this complex plays a role in human airway epithelial proliferation. The integrin receptors are composed of alpha and beta subunits and each subunit crosses the membrane once, with most of the polypeptide residing in the extracellular space, and has two short cytoplasmic domains. Integrin beta-8 is a single-pass type I membrane protein that associates with alpha V chain in a non-covalent manner to make a heterodimer that serves as a receptor for vitronectin, fibrin, and the latency associated peptide (LAP). Integrin beta-8 binding to LAP is reported to promote the proteolytic release of active TGF-beta from LAP. Integrin beta-8 contains a VWFA domain and four cysteine-rich repeats. It is required for vascular morphogenesis in the embryonic brain and yolk sac. Higher expression of integrin beta-8 is observed in placenta, kidney, brain, ovary, uterus, and in several transformed cells. It is transiently expressed in 293 human embryonic kidney cells. Higher expression of ITGB8 is linked to high angiogenic and poorly invasive glioblastoma tumors. Conversely, low expression of ITGB8 correlates with highly invasive, but low angiogenic tumors. Higher expression of integrin beta-8 is observed in placenta, kidney, brain, ovary, uterus, and in several transformed cells. It is transiently expressed in 293 human embryonic kidney cells.

Specificity

Clone11D4 specifically detects isoform 2 of human integrin beta-8.

Immunogen

A truncated recombinant human Integrin B8, isoform 2, corresponding to amino acids 136-769.

Application

Research Category
Cell Structure
This mouse monoclonal Anti-Integrin B8 Antibody, clone11D4, Cat. No. MABT912 is tested for use in Western Blotting for the detection of Integrin B8.

Quality

Evaluated by Western Blotting in HT-29 cell lysate.

Western Blotting Analysis: 1 µg/mL of this antibody detected Integrin B8 in 10 µg of HT-29 cell lysate.

Target description

~71 kDa observed; 71.28 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service