Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABT900

Sigma-Aldrich

Anti-Connexin 45 Antibody, clone P3C9

clone P3C9, from mouse

Synonym(s):

Gap junction gamma-1 protein, Cx45, Gap junction alpha-7 protein

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$484.00

$484.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
$484.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$484.00


Check Cart for Availability

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

P3C9, monoclonal

species reactivity

rat, mouse

species reactivity (predicted by homology)

rhesus macaque (based on 100% sequence homology), porcine (based on 100% sequence homology), bovine (based on 100% sequence homology), human (based on 100% sequence homology), rabbit (based on 100% sequence homology), canine (based on 100% sequence homology), chicken (based on 100% sequence homology)

technique(s)

western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

bovine ... Gjc1(511119)
dog ... Gjc1(490936)
human ... GJC1(10052)
mouse ... Gjc1(14615)
pig ... Gjc1(100048959)
rat ... Gjc1(266706)
rhesus monkey ... Gjc1(715463)

General description

Gap junction gamma-1 protein (UniProt: A4GG66; also known as Connexin-45, Cx45, Gap junction alpha-7 protein) is encoded by the Gjc1 (also known as Gja7) gene (Gene ID: 266706) in rat. The gap junction channels are dodecamers of transmembrane proteins of the connexin family. Connexins usually have a short half-life ( 1.5 - 5 h) and this short half-life is a key element of coupling regulation as it allows very dynamic and acute changes in gap junction regulation and gap junctional communication. In cardiomyocytes three types of connexins have been identified: Connexin 40, 43, and 45. Each of these connexins form channels with unique and specific electrophysiological properties. Connexin-45 is a multi-pass membrane protein with ubiquitous expression. It consists of a cluster of closely packed pairs of transmembrane channels, the connexons, through which materials of low molecular weights diffuse from one cell to a neighboring cell. Connexin 45 is the first cardiac connexin expressed during embryonic development and plays an important role in propagating the action potential from the conduction system to the working myocardium. Connexin 45 gap junction channels are reported to close when the membrane potential becomes negative. This prevents retrograde conduction from myocardium to the conduction system.

Specificity

Clone clone P3C9 detects Connexin-45 in rat and mouse cells. It targets an epitope within 14 amino acids from the C-terminal half of rat connexin 45.

Immunogen

KLH-conjugated linear peptide corresponding to 14 amino acids from the C-terminal half of rat connexin 45.

Application

Research Category
Cell Structure
This mouse monoclonal Anti-Connexin 45, clone P3C9, Cat. No. MABT900 is tested for use in Western Blotting for the detection of Connexin-45.
Western Blotting Analysis: A representative lot detected Connexin-45 in Western Blotting applications (Manias, J.L., et. al. (2008). Cardiovasc Res. 80(3):385-95).

Western Blotting Analysis: A representative lot detected Connexin-45 in Western Blotting applications (Kopanic, J.L., et. al. (2013). Front Pharmacol. 4:106).

Quality

Evaluated by Western Blotting in mouse heart tissue lysate.

Western Blotting Analysis: 1 µg/mL of this antibody detected Connexin 45 in 10 µg of mouse heart tissue lysate.

Target description

~45 kDa observed; 45.67 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG2b in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service