Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

MABT493

Sigma-Aldrich

Anti-IL-34, clone 1D12 Antibody

ascites fluid, clone 1D12, from mouse

Synonym(s):

Interleukin-34, IL-34

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$388.00

$388.00


Check Cart for Availability

Request a Bulk Order
Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
100 μL
$388.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$388.00


Check Cart for Availability

Request a Bulk Order
Notify Me

Get notified when this item is ready to ship via email.

biological source

mouse

Quality Level

antibody form

ascites fluid

antibody product type

primary antibodies

clone

1D12, monoclonal

species reactivity

human

technique(s)

flow cytometry: suitable
immunofluorescence: suitable

isotype

IgG1

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... IL34(146433)

Related Categories

General description

Intereukin 34, also known as IL-34 or IL34 is a cytokine that promotes the differentiation and viability of moncytes and macrophages. IL-34 stimulate the phosphorylation of MAPL1/ERK2/ERK2 and MAPK3/ERK1. IL-34 is a ligand for colony-stimulating factor CSF1R. IL-34 is predominantly expressed in spleen, and also detected in a range of other tissues including heart, liver, kidney, thymus, testis, ovary, small intestine, prostate and colon.

Immunogen

Purified recombinant fragment of human IL-34 expressed in E. Coli

Application

Anti-IL-34, clone 1D12 Antibody is an antibody against IL-34 for use in flow cytometry & immunofluorescence.
Immunofluorescence Analysis: A 1:200-1,000 dilution from a representative lot detected IL-34 in U251 cells.
Research Category
Cell Structure
Research Sub Category
Cytoskeleton

Quality

Flow Cytometry Analysis: A 1:200-400 dilution from a representative lot detected IL-34 in K562 cells.

Target description

27 kDa calculated

Physical form

Mouse monoclonal IgG1 ascitic fluid containing up to 0.1% sodium azide.
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Muhammad Baghdadi et al.
Inflammation and regeneration, 38, 12-12 (2018-07-14)
Cysts of the salivary glands are common lesions that occur in the context of various etiologies. Although the diagnostic importance of cysts in salivary gland diseases has been well studied, molecular mechanisms that control the related pathological process remain largely
Nabeel Kajihara et al.
Cancer immunology, immunotherapy : CII, 72(4), 851-864 (2022-09-15)
Triple-negative breast cancer (TNBC) is an aggressive breast cancer subtype characterized by a lack of therapeutic targets. The paucity of effective treatment options motivated a number of studies to tackle this problem. Immunosuppressive cells infiltrated into the tumor microenvironment (TME)
Nanumi Han et al.
Inflammation and regeneration, 38, 3-3 (2018-03-09)
Immunotherapies that target immune-checkpoint molecules such PD-1 have helped to achieve durable responses in melanoma treatment. However, 25% of melanoma patients who showed objective responses to PD-1 blockade develop resistance and suffer from disease progression and ultimately death, which necessitates
Nanumi Han et al.
STAR protocols, 2(2), 100460-100460 (2021-04-27)
Previously, we identified a therapy-resistant role of IL-34 in an immune checkpoint blockade in murine models. To investigate whether a similar mechanism is applicable in human tumors as well, we used this protocol for the selection of IL-34-neutralizing antibody and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service