Breast cancer anti-estrogen resistance protein 3 (BCAR3), also known as Novel SH2-containing protein 2 (NSP2) and SH2 domain-containing protein 3B (SH2D3B) is an adapter protein that regulates the proliferation in breast cancer cells. BCAR3 may also be regulated by cellular adhesion to extracellular matrix proteins. BCAR3 is found in several cancer cell lines; however, it is not found in nonmalignant breast tissue. When BCAR3 is overexpressed, it confers anti-estrogen resistance in breast cancer cell lines.
Immunogen
GST-tagged recombinant protein corresponding to human BCAR3.
Application
Anti-BCAR3, clone 9D9.1 is an antibody against BCAR3 for use in Western Blotting.
Research Category Cell Structure
Research Sub Category Adhesion (CAMs)
Quality
Evaluated by Western Blotting in U2OS cell lysate.
Western Blotting Analysis: 0.5 µg/mL of this antibody detected BCAR3 in 10 µg of U2OS cell lysate.
Target description
~93 kDa observed
Physical form
Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Storage and Stability
Stable for 1 year at 2-8°C from date of receipt.
Other Notes
Concentration: Please refer to lot specific datasheet.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Not finding the right product?
Try our Product Selector Tool.
Storage Class
12 - Non Combustible Liquids
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.