Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

MABT177

Sigma-Aldrich

Anti-Syndecan-1 Antibody, clone 11G2.1

clone 11G2.1, from mouse

Synonym(s):

Syndecan-1, CD138, SYND1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

11G2.1, monoclonal

species reactivity

human

technique(s)

flow cytometry: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... SDC1(6382)

General description

Syndecan-1 (UniProt P18827; also known as CD138, SYND1) is encoded by the SDC1 (also known as SDC) gene (Gene ID 6382) in human. Syndecan-1 belongs to a family of transmembrane heparan sulfate proteoglycans (HSPGs), members of which also include syndecan-2 (fibroglycan, SDC2), syndecan-3 (N-syndecan, SDC3), and syndecan-4 (amphiglycan, ryudocan, SDC4). Syndecan-1 is produced with a signal peptide (a.a. 1-22), the removal of which yields the mature protein with a large extracellular region (a.a. 23-254), a transmembrane segment (a.a. 255-275), and a cytoplasmic tail (a.a. 276-310). Syndecan-1 is modified at its extracellular domain on a.a. 45 and 47 with O-linked heparan sulfate (HS) that plays an important role in mediating its interaction with HS-binding protein, including growth factors (e.g. bFGF/FGF2, PDGF, TGF-β, and VEGF) and extracellular matrix proteins (e.g. collagens, fibronectin, thrombospondin, and tenascin). Syndecan-1 thus funtions as a coreceptor for growth factor receptors and acts as an anchor connecting the extracellular matrix to the intracellular cytoskeleton. Syndecan extracellular domain can be proteolytically cleaved off at a juxtamembrane site and upregulated shedding of the ectodomain occurs in response to growth factors, chemokines, heparanase, microbial toxins, insulin, and cellular stress. SDC1 deregulation contributes to cancer progression by promoting angiogenesis and cancer cell proliferation, metastasis, and invasion.

Specificity

Clone 11G2.1 recognizes an extracellular epitope.

Immunogen

Epitope: Extracellular domain.
GST-tagged recombinant human Syndecan-1 extracellular domain fragment.

Application

Anti-Syndecan-1 Antibody, clone 11G2.1 is an antibody against Syndecan-1 for use in Western Blotting, Flow Cytometry.
Flow Cytometry Analysis: 1.0 µg from a representative lot detected Syndecan-1 in one million Raji and Ramos cells.
Research Category
Cell Structure
Research Sub Category
Adhesion (CAMs)

Quality

Evaluated by Western Blotting of human syndecan-1 GST fusion protein.

Western Blotting Analysis: 0.1 µg/mL of this antibody detected 0.05 µg of a GST-tagged human syndecan-1 extracellular domain fragment.

Target description

30.00 kDa calculated. Uncharacterized band(s) may appear in some lysates.

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service