Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABT175

Sigma-Aldrich

Anti-ADAMTS5 Antibody, clone 3A2.1

clone 3A2.1, from mouse

Synonym(s):

A disintegrin and metalloproteinase with thrombospondin motifs 5, ADAM-TS 5, ADAM-TS5, ADAMTS-5, EC 3.4.24.-, A disintegrin and metalloproteinase with thrombospondin motifs 11, ADAM-TS 11, ADAMTS-11, ADMP-2, Aggrecanase-2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

antibody form

purified antibody

antibody product type

primary antibodies

clone

3A2.1, monoclonal

species reactivity

human

technique(s)

immunohistochemistry: suitable (paraffin)

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ADAMTS5(11096)

General description

The ADAMTS (A Disintegrin and Metalloprotease with Thrombospondin motifs) family of secreted proteases comprises at least 20 members in mammals, and several members in invertebrates. ADAMTS members resemble the membrane-bound ADAM family of proteases in the presence of a prodomain, a catalytic domain and a disintegrin-like domain. However, ADAMTS members lack a transmembrane domain, and instead contain varying numbers of thrombospondin type I modules. ADAMTS-5, along with ADAMTS-4, cleaves aggrecan between Glu373 and Ala374, a physiological cleavage site in arthritic cartilage. In addition, expression of ADAMTS-5 is elevated in human chondrocytes treated with inflammatory cytokine interleukin-1, but is constitutively expressed in bovine cartilage explants. 70 and 53 kD forms of ADAMTS-5 have been shown to be present at varying ratios in fibroblast-like synoviocytes and extracts of rheumatoid arthritis, osteoarthritis and normal synovial tissue.

Immunogen

Epitope: C-terminus
GST-tagged recombinant protein corresponding to the C-terminus of human ADAMTS5.

Application

Detect ADAMTS5 using this Anti-ADAMTS5 Antibody, clone 3A2.1 validated for use in IHC (Paraffin).
Research Category
Cell Structure
Research Sub Category
Cytoskeletal Signaling

Quality

Evaluated by Immunohistochemistry in human thalamus tissue.

Imunnohistochemistry Analysis: A 1:50 dilution of this antibody detected ADAMTS5 in human thalamus tissue.

Target description

101.7 kDa calculated

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service