Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

MABT157

Sigma-Aldrich

Anti-Parvin alpha Antibody, clone 7E2.1

clone 7E2.1, from mouse

Synonym(s):

Alpha-parvin, α-Parvin, a-Parvin, topaxin, CH-ILKBP, Calponin-like integrin-linked kinase-binding protein, Matrix-remodeling-associated protein 2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$333.00

$333.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
$333.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$333.00


Check Cart for Availability

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

7E2.1, monoclonal

species reactivity

human

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... PARVA(55742)

General description

The protein named α-Parvin or Parvin, or Actopaxin, or Calponin-like integrin-linked kinase-binding protein (CH-ILKBP), or Matrix-remodeling-associated protein 2 (MXRA2), and encoded by the gene PARVA/MXRA2, is a critical adhesion coupling protein involved in many systems including sarcomere organization, smooth muscle contraction, cardiovascular development, angiogenesis, actin cytoskeleton organization and the establishment of cell polarity, adhesion, spreading and migration. α-Parvin is required for the adhesion of smooth muscle cell to endothelial cells during blood vessel development and for the formation of lamellipodia and ciliogenesis. α-Parvin is localized to the cell junction and focal adhesions. The protein is peripheral membrane protein located on the cytoplasmic side of the cell membranes and interacts with the cytoskeleton in the cytoplasm. In muscle cells it is associated with the Z line of the sarcomere. α-Parvin is expressed many tissues including muscle, kidney and liver.

Immunogen

GST-tagged recombinant protein corresponding to human Parvin.

Application

Immunohistochemistry Analysis: A 1:2,000 dilution and a 1:50 dilution from a representative lot detected Parvin alpha in human kidney and skeletal muscle tissues, respectively.
This Anti-Parvin alpha Antibody, clone 7E2.1 is validated for use in western blotting & IHC (Paraffin) for the detection of Parvin alpha.

Quality

Evaluated by Western Blotting in human heart tissue.

Western Blotting Analysis: 0.2 µg/mL of this antibody detected Parvin alpha in 200 µg of human heart tissue.

Target description

~42 kDa observed

Physical form

Format: Purified

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Haibin Yang et al.
eLife, 10 (2021-02-16)
Communications between actin filaments and integrin-mediated focal adhesion (FA) are crucial for cell adhesion and migration. As a core platform to organize FA proteins, the tripartite ILK/PINCH/Parvin (IPP) complex interacts with actin filaments to regulate the cytoskeleton-FA crosstalk. Rsu1, a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service