Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABT123

Sigma-Aldrich

Anti-Pirh2 Antibody, clone 11G8.1

clone 11G8.1, from mouse

Synonym(s):

RING finger and CHY zinc finger domain-containing protein 1, Androgen receptor N-terminal-interacting protein, CH-rich-interacting match with PLAG1, E3 ubiquitin-protein ligase Pirh2, RING finger protein 199, Zinc finger protein 363, p53-induced RING-H2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$294.00

$294.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
$294.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$294.00


Check Cart for Availability

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

11G8.1, monoclonal

species reactivity

human, mouse

technique(s)

western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... RCHY1(25898)

General description

Pirh2, also known as Androgen receptor N-terminal-interacting protein or CH-rich-interacting match with PLAG1 or E3 ubiquitin-protein ligase Pirh2 or RING finger protein 199 or Zinc finger protein 363 or p53-induced RING-H2 protein (Pirh2), encoded by the gene RCHY1/ARNIP/CHIMP/PIRH2/RNF199/ZNF363, is an E3 ubiquitin ligase protein that mediates the E3-dependent ubiquitination and proteasomal degradation of a number of important signaling and DNA methylation proteins including p53/TP53, p73, HDAC1 and CDKN1B. Pirh2 also monoubiquitinates the translesion DNA polymerase POLH and contributes to the regulation of the cell cycle progression. Additionally, Pirh2 increases androgen receptor transcription factor activity as well and Interacts with AR, p53/TP53, MDM2, HDAC1, KAT5, PLAG1, PLAGL2, CDKN1B, COPE, UBE2D2 and GORAB/NTKLBP1 proteins. Pirh2 is localized to the cytoplasm and nucleus, and can be found in nucleus speckles. Pirh2 is expressed in all cell types examined and is up-regulated during the S phase of the cell cycle. Pirh2 is expressed at low levels during G phase.

Immunogen

GST-tagged recombinant protein corresponding to human Pirh2.

Application

Anti-Pirh2 Antibody, clone 11G8.1 is a highly specific mouse monoclonal antibody & that targets Pirh2 & has been tested in western blotting.
Research Category
Cell Structure
Research Sub Category
ECM Proteins

Quality

Evaluated by Western Blotting in H1299 cell lysate.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected Pirh2 in 10 µg of H1299 cell lysate.

Target description

~30 kDa observed

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG2aκ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service