Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABS826

Sigma-Aldrich

Anti-Rsk1 Antibody, clone 3G5.1

clone 3G5.1, from mouse

Synonym(s):

Ribosomal protein S6 kinase alpha-1, 90 kDa ribosomal protein S6 kinase 1, MAP kinase-activated protein kinase 1a, MAPK-activated protein kinase 1a, MAPKAP kinase 1a, MAPKAPK-1a, p90-RSK 1, p90RSK1, p90S6K, Ribosomal S6 kinase 1, RSK-1, S6K-alpha-1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$457.00

$457.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
$457.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$457.00


Check Cart for Availability

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

3G5.1, monoclonal

species reactivity

rat

species reactivity (predicted by homology)

mouse (based on 100% sequence homology), Xenopus (based on 100% sequence homology), human (based on 100% sequence homology), porcine (based on 100% sequence homology), chicken (based on 100% sequence homology), bovine (based on 100% sequence homology)

technique(s)

western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... RPS6KA1(6195)

General description

Ribosomal protein S6 kinase alpha-1 (EC 2.7.11.1; UniProt Q15418; also known as 90 kDa ribosomal protein S6 kinase 1, MAP kinase-activated protein kinase 1a, MAPK-activated protein kinase 1a, MAPKAP kinase 1a, MAPKAPK-1a, p90-RSK 1, p90RSK1, p90S6K, Ribosomal S6 kinase 1, RSK-1, S6K-alpha-1) is encoded by the RPS6KA1 (also known as MAPKAPK1A, RSK1) gene (Gene ID 6195) in human. Ribosomal s6 kinases (RSKs) are serine/threonine kinases that play a role in mediating Mek1/2-Erk1/2 mitogen-activated protein (MAP) kinase pathway signaling events. There exist two types of RSKs, p70rsk (a.k.a. S6 Kinase or S6-H1 kinase) and p90rsk (a.k.a. MAPK-activated protein kinase). Six p90rsk-coding genes have been identified in human, RPS6KA1 (MAPKAPK1A, RSK1), RPS6KA2 (MAPKAPK1C, RSK3), RPS6KA3 (MAPKAPK1B, RSK2), RPS6KA4 (MSK2), RPS6KA5 (MSK1), and RPS6KA6 (RSK4). p90RSK1 is activated by sequential phosphorylation events, where it is first phosphorylated by Erk1/2 on the C-terminal kinase domain (CTKD) Thr573, leading to subsequent Ser380 phosphorylation in the N-terminal AGC kinase-type domain. Alternatively, Ser380 can be phosphorylated directly by MK2/3 independent of CTKD Thr573 phosphorylation.

Specificity

Clone 3G5.1 target epitope sequence is present in all four human Rsk1 spliced isoforms reported by UniProt (Q15418).

Immunogen

Epitope: Linker region between two kinase domains.
KLH-conjugated linear peptide corresponding to a sequence from the linker region between the two human Rsk1 kinase domains.

Application

Anti-Rsk1 Antibody, clone 3G5.1 is an antibody against Rsk1 for use in Western Blotting.

Quality

Evaluated by Western Blotting in PC12 cell lysate.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected Rsk1 in 10 µg of PC12 rat pheochromocytoma cell lysate.

Target description

~85 kDa observed. 82.88 kDa (rat), 86.47 kDa (mouse), 82.72 kDa (human isoform 1), 83.93 kDa (human isoform 2), 72.70 kDa (human isoform 3), and 81.15 kDa (human isoform 4) calculated. Uncharacterized band(s) may appear in some lysates.

Physical form

Format: Purified

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service