Skip to Content
MilliporeSigma
All Photos(5)

Key Documents

MABS777

Sigma-Aldrich

Anti-FADS1, clone EPR6898, Rabbit Monoclonal Antibody

clone EPR6898, from rabbit

Synonym(s):

Fatty acid desaturase 1, Delta(5) fatty acid desaturase, D5D, Delta(5) desaturase, Delta-5 desaturase, FADS1, FADSD5

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

antibody form

unpurified

antibody product type

primary antibodies

clone

EPR6898, monoclonal

species reactivity

human

technique(s)

flow cytometry: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... FADS1(3992)

General description

Fatty acid desaturase 1, also known as Delta (5 fatty acid desaturase or D5D or Delta (5 desaturase or Delta-5 desaturase, and encoded by the gene name FADS1 or FADSD5, is a member of the fatty acid desaturase (FADS) family. Desaturase enzymes regulate unsaturation of fatty acids through the introduction of double bonds between defined carbons of the fatty acyl chain. FADS family members are considered fusion products composed of an N-terminal cytochrome b5-like domain and a C-terminal multiple membrane-spanning desaturase portion, both of which are characterized by conserved histidine motifs.

Specificity

Dissociation constant (KD) is 7.20 x 10-11 M

Immunogen

Synthetic peptide corresponding to human FADS1.

Application

Anti-FADS1, clone EPR6898, Rabbit Monoclonal Antibody is an antibody against FADS1 for use in Western Blotting, Immunohistochemistry, Flow Cytometry, Immunocytochemistry.
Immunohistochemistry Analysis: A 1:100-250 dilution from a representative lot detected FADS1 in human heart tissue.

Flow Cytometry Analysis: A 1:100-1,000 dilution from a representative lot detected FADS1 in HepG2 cells.

Immunocytochemistry Analysis: A 1:100-250 dilution from a representative lot detected FADS1 in A549 cells.

Produced in collaboration with Epitomics
Research Category
Signaling
Research Sub Category
Lipid Signaling

Quality

Evaluated by Western Blotting in human fetal lung, human fetal liver, human fetal brain, and HepG2 lysates.

Western Blotting Analysis: A 1:1,000-10,000 dilution of this antibody detected FADS1 in human fetal lung, human fetal liver, human fetal brain, and HepG2 lysates.

Target description

~52 kDa observed. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Unpurified
Rabbit monoclonal in buffer containing PBS with up to 0.1% sodium azide, 0.05% BSA, and 50% glycerol.
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Note: Variability in freezer temperatures below -20°C may cause glycerol containing solutions to become frozen during storage.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service