Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

MABS501

Sigma-Aldrich

Anti-Anoctamin-5 Antibody, clone 5F7

clone 5F7, from mouse

Synonym(s):

Anoctamin-5, Gnathodiaphyseal dysplasia 1 protein, Transmembrane protein 16E

Sign Into View Organizational & Contract Pricing

Select a Size

200 μL
$352.00

$352.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
200 μL
$352.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$352.00


Check Cart for Availability

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

unpurified

antibody product type

primary antibodies

clone

5F7, monoclonal

species reactivity

human, mouse

species reactivity (predicted by homology)

rat (based on 100% sequence homology)

technique(s)

western blot: suitable

isotype

IgMκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ANO5(203859)

General description

Anoctamin-5 (UniProt Q75V66; also known as Gnathodiaphyseal dysplasia 1 protein, Integral membrane protein GDD1, Transmembrane protein 16E) is encoded by the ANO5 (also known as GDD1, LGMD2L, TMEM16E) gene (Gene ID 203859) in human. The human Anoctamins (ANO) compose a family of at least ten proteins, all exhibiting eight transmembrane domains and a DUF590 domain of unknown function. The ANO1, ANO2, ANO6, ANO8, and ANO9 are characterized to code for the elusive calcium-activated chloride channels (CaCCs). While the physiological function of anoctamin-5 is not yet characterized, dominant mutations in ANO5 have been found in the rare skeletal disorder gnathodiaphyseal dysplasia. In addition, recessive mutations in ANO5 are found to cause proximal muscular dystrophy (LGMD2L) and distal Miyoshi myopathy (MMD3).

Immunogen

Epitope: Near C-terminus
Linear peptide corresponding to mouse Anoctamin-5 near the C-terminus.

Application

Research Category
Signaling
Research Sub Category
Signaling Neuroscience
This Anti-Anoctamin-5 Antibody, clone 5F7 is validated for use in Western Blotting for the detection of Anoctamin-5.
Western Blotting Analysis: A 1:100 dilution from a representative lot detected Anoctamin-5 in 10 µg of mouse leg muscle tissue lysate.

Quality

Evaluated by Western Blotting in mouse skeletal muscle tissue lysate.

Western Blotting Analysis: A 1:500 dilution of this antibody detected Anoctamin-5 in 10 µg of mouse skeletal muscle tissue lysate.

Target description

~37 kDa observed. ~37 kDa observed. Although the full-length protein has a calculated molecular weight of 107.2 kDa (Uniprot Q75V66), only a ~37 kDa band corresponding to a short spliced variant can be detected by Western blotting when using muscle tissue lysates. A band corresponding to the full-size protein and a 55 kDa band can be detected in addition to the 37 kDa band when using lysates prepared from human embryonal rhabdomyosarcoma (RMS) RD cells, C2C12 murine myoblasts, and fibroblasts.

Physical form

Format: Unpurified
Mouse monoclonal IgMκ in buffer containing supernatant without preservatives.
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service