Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

MABS475

Sigma-Aldrich

Anti-GRP-78 Antibody, clone C107

clone C107, from mouse

Synonym(s):

78 kDa glucose-regulated protein, GRP-78, Endoplasmic reticulum lumenal Ca(2+)-binding protein grp78, Heat shock 70 kDa protein 5, Immunoglobulin heavy chain-binding protein, BiP

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

C107, monoclonal

species reactivity

mouse, human

technique(s)

flow cytometry: suitable
western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... HSPA5(3309)

General description

The gene HSPA5 encodes a protein named GRP-78 (78 kDa glucose-regulated protein) also known as BiP (immunoglobulin heavy chain-binding protein) or heat shock 70 kDa protein 5. While originally discovered as one of set of genes that were induced upon cellular starvation and stress, HSPA5 encoded GRP-78 is now known to be an important member of the heat-shock family of proteins. The HSPA5 encoded GRP-78 protein appears to play a role in helping in the assembly of multimeric protein complexes in the endoplasmic reticulum (ER). Cellular localization is in the endoplasmic reticulum lumen and appears as cytoplasmic-like staining. The HSPA5 encoded protein is an essential component of the translocation processes within the endoplasmic reticulum and also functions in the retrograde transport across the ER membrane of incorrectly folded proteins destined to be degraded by the ubiquitin proteasome system. The HSPA5 encoded protein, while widely expressed in all cells types under normal conditions, is particularly induced under conditions of cellular stress, heat, or toxins and conditions which lead to the accumulation of unfolded proteins in the ER.

Immunogen

Recombinant protein corresponding to human GRP-78.

Application

Research Category
Signaling
Research Sub Category
Insulin/Energy Signaling
This Anti-GRP-78 Antibody, clone C107 is validated for use in western blotting, flow cytometry & Inhibits activity/Function for the detection of GRP-78.
Western Blotting Analysis: 1 µg/mL from a representative lot detected GRP-78 in 10 µg of HeLa cell lysate.

Flow Cytometry Analysis: A representative lot from an independent laboratory detected GRP-78 in B16F1 cells from tumor bearing mice (de Ridder, G. G., et al. (2012). Melanoma Res. 22(3):225-235.).

Activity Assay Analysis: A representative lot from an independent laboratory reduces tumor growth in mice with B16F1 melanoma cells (de Ridder, G. G., et al. (2012). Melanoma Res. 22(3):225-235.).

Quality

Evaluated by Western Blotting in HepG2 cell lysate.

Western Blotting Analysis: 1 µg/mL of this antibody detected GRP-78 in 10 µg of HepG2 cell lysate.

Target description

~72 kDa observed

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG2bκ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service