Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

MABS458

Sigma-Aldrich

Anti-Betacellulin Antibody, clone 6G10.1

clone 6G10.1, from mouse

Synonym(s):

Probetacellulin, Betacellulin, BTC

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$352.00

$352.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
$352.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$352.00


Check Cart for Availability

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

6G10.1, monoclonal

species reactivity

human

technique(s)

immunohistochemistry: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... BTC(685)

General description

The human BTC gene (Gene ID 685) encodes the 178 aa preprobetacellulin (UniProt P35070) protein. Post-translatioinal removal of the signal peptide (aa 1-31) produces probetacellulin (aa 32-178), which further matures into the 80 aa betacellulin (aa 32-111) following propeptide (aa 112-178) removal. Betacellulin is an EGF family growth factor with a higher affinity and specificity for ErbB1/EGFR and ErbB4. Betacellulin plays an important role in the development of the acute exacerbations of chronic obstructive pulmonary disease. Betacellulin is also reported to induce CXCL8 (IL-8) production from human lung cancer cells through EGFR–PI3K–Akt–ERK pathway activation, resulting in the recruitment of inflammatory cells in the cancer tissue and creating an inflammatory tumor microenvironment.

Specificity

Clone 6G10.1 is expected to detect prepro-, pro-, and mature betacellulin.

Immunogen

Epitope: Mature betacellulin
GST-tagged human recombinant full-length mature betacellulin.

Application

Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected Betacellulin in human pancreas tissue.
Research Category
Signaling
Research Sub Category
Signaling Neuroscience
This Anti-Betacellulin Antibody, clone 6G10.1 is validated for use in Western Blotting, Immunohistochemistry for the detection of Betacellulin .

Quality

Evaluated by Western Blotting in human pancreas tissue lysate.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected Betacellulin in 10 µg of human pancreas tissue lysate.

Target description

~20 kDa observed. Preprobetacellulin (19.7 kDa calculated); Probetacellulin (16.6 kDa calculated); Betacellulin (9.0 kDa calculated). Due to posttranslational glycosylation, target band(s) will migrate with a higher apparent M.W. than caluclated.

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service