Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABS2045

Sigma-Aldrich

Anti-HPa2 Antibody, clone DHIC2-2B4

clone DHIC2-2B4, from mouse

Synonym(s):

Human pancreas alpha cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

antibody form

purified antibody

antibody product type

primary antibodies

clone

DHIC2-2B4, monoclonal

species reactivity

human

packaging

antibody small pack of 25 μg

technique(s)

flow cytometry: suitable
immunohistochemistry: suitable (paraffin)

isotype

IgG1λ

target post-translational modification

unmodified

Related Categories

General description

Pancreas performs both the endocrine and the exocrine function. The bulk of pancreas is composed of exocrine cells that produce digestion enzymes which are released into ducts to be emptied into the duodenum. The endocrine pancreas is composed of islets of Langerhans that produce hormones. At least five distinct endocrine cell types have been identified in the endocrine pancreas, each producing a different hormone. Beta cells produce insulin, which is considered as an anabolic hormone and regulates blood glucose levels and is also involved in fat and protein synthesis. Alpha cells produce glucagon that acts when blood glucose levels are low. Ductal cells of pancreas form the epithelial lining of the branched tubes that deliver digestive enzymes into the duodenum. They also secrete bicarbonate to neutralize acidic contents coming in from stomach. Duct cells may contribute up to 10% of all pancreatic cells. Acinar cells function as a unit of the exocrine pancreas and they synthesize, store, and secrete digestive enzymes (lipase, alpha amylase, trypsin, and chymotrypsin), which under normal physiological conditions are activated only after reaching the duodenum. Premature activation of these enzymes within pancreatic acinar cells can lead to acute pancreatitis. Clone DHIC2-2B4 can be used to detect alpha cells in human pancreatic islets. (Ref.: Dorrell, C., et al. (2008). Stem Cell Research (2008). 1(3); 183-194; Grapin-Botton, A (2005). Int. J. Biochem. Cell Biol. 37(3); 504-510; Leung, PS (2006). Int. J. Biochem. Cell Biol. 38(7); 1024-1030).

Specificity

Clone DHIC2-2B4 specifically detects alpha cells in human pancreatic tissue sections.

Immunogen

Trypsin-dissociated human pancreatic islet cells containing low levels of exocrine and ductal cells.

Application

Detects Alpha cells of human pancreatic islets using this mouse monoclonal Anti-HPa2, clone DHIC2-2B4, Cat. No. MABS2045, tested for use in Immunohistochemistry (Paraffin) and Flow Cytometry.
Immunohistochemistry Analysis: A representative lot detected HPa2 in Immunohistochemistry applications (Dorrell, C., et. al. (2008). Stem Cell Res. 1(3):183-94).

Flow Cytometry Analysis: A representative lot detected HPa2 in Flow Cytometry applications (Dorrell, C., et. al. (2008). Stem Cell Res. 1(3):183-94; Bramswig, N.C., et. al. (2013). J Clin Invest. 123(3):1275-84; Dorrell, C., et. al. (2016). Nat Commun. 7:11756).

Quality

Evaluated by Immunohistochemistry (Paraffin) in alpha cells of human pancreatic tissue sections.

Immunohistochemistry (Paraffin) Analysis: A 1:50 dilution of this antibody detected HPa2 in alpha cells of human pancreatic tissue sections.

Physical form

Format: Purified

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service