Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABS199

Sigma-Aldrich

Anti-Linear Ubiquitin, clone 1E3 Antibody

clone 1E3, from rabbit

Synonym(s):

Polyubiquitin-C, Ubiquitin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

1E3, monoclonal

species reactivity

human

technique(s)

immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgGκ

UniProt accession no.

shipped in

wet ice

Gene Information

human ... UBC(7316)

General description

Ubiqutin (Ub) is initially produced as a 229-a.a. Polyubiquitin-B (UniProt P0CG47) precursor protein encoded by the UBB gene (Gene ID 7314) or a 685-a.a. Polyubiquitin-C precursor protein (UniProt P0CG48) encoded by the UBC gene (Gene ID 7316) in human. The C-termini of Polyubiquitin-B (Cys229) and Polyubiquitin-C (Val685) are post-translationally removed, followed by additional posttranslational cleavages to yield multiple copies of idential 76-amino acid ubiquitin (also known as high mobility group nonhistone protein HMG-20) molecules (three copies per Polyubiquitin-B and nine copiesf per Polyubiquitin-C). Ub is involved in one of the most common post-translational modifications (PTMs) of cellular proteins, where Ub is linked covalently via its carboxyl terminus (Gly76) to usually Lys residues in target proteins. A given target Lys residue can be linked to one single Ub molecule (monoubiquitylated) or to a chain of Ub molecules (polyubiquitylated). In a polyUb chain, Ub molecules can be linked through one of the seven Lys residues (K6, K11, K27, K29, K33, K48, and K63) or through the Ub N-terminus Met1 residue (which generates linear chains). An Ub chain with a single linkage type is referred to as homotypic, whereas an heterotypic chain contains mixed linkages within the same chain, which can lead to either branched/forked (as a result of one Ub linked to two or more Ubs) or non-branched Ub chains. Lys48-linked chains mark proteins for proteasomal degradation, while Lys63-linked chains regulate the activity/function of cellular proteins.

Immunogen

Recombinant protein corresponding to all linear ubiquitin linkages.

Application

Immunocytochemistry Analysis: A 1:50 dilution from a representative lot detected linear ubiquitin in Hoi-1L/Hoip transfected Hela cells.

Immunoprecipitatation Analysis: 10 µg from a representative lot immunoprecipitated linear ubiquitin from 500 µg of Hoi-1L/Hoip transfected HEK293 cells.

Western Blotting Analysis: A representative lot from an independent laboratory detected K64-linked diUbiquitin (Matsumoto, M. L., et al. (2012). J Mol Biol. 418(3-4):134-144.).
Research Category
Signaling
Research Sub Category
Cell Cycle, DNA Replication & Repair
This Anti-Ubiquitin antibody, clone 1E3 is validated for use in western blotting, IP & ICC for the detection of Linear Ubiquitin.

Quality

Evaluated by Western Blotting in linear polyubiquitin chains, K48 Ubiquitin 2-7, K63 Ubiquitin 2-7, and K11-only Ubn-ubiquitinylated HM substrate protein synthesized using K11-only mutant ubiquitin.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected linear ubiquitin chains, and not K48 Ubiquitin 2-7, K63 Ubiquitin 2-7, or K11-only Ubn-ubiquitinylated HM substrate protein synthesized using K11-only mutant ubiquitin.

Target description

Multiple molecular weight bands observed

Physical form

Format: Purified
Protein A purified
Purified rabbit monoclonal IgGκ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aurelia Stangl et al.
Nature communications, 10(1), 4320-4320 (2019-09-22)
OTULIN (OTU Deubiquitinase With Linear Linkage Specificity) specifically hydrolyzes methionine1 (Met1)-linked ubiquitin chains conjugated by LUBAC (linear ubiquitin chain assembly complex). Here we report on the mass spectrometric identification of the OTULIN interactor SNX27 (sorting nexin 27), an adaptor of the
Arun K Haldar et al.
mBio, 7(6) (2016-12-15)
The cytokine gamma interferon (IFN-γ) induces cell-autonomous immunity to combat infections with intracellular pathogens, such as the bacterium Chlamydia trachomatis The present study demonstrates that IFN-γ-primed human cells ubiquitinate and eliminate intracellular Chlamydia-containing vacuoles, so-called inclusions. We previously described how
Ran Wei et al.
Genes & development, 31(11), 1162-1176 (2017-07-14)
Stimulation of cells with TNFα leads to the formation of the TNF-R1 signaling complex (TNF-RSC) to mediate downstream cellular fate decision. Activation of the TNF-RSC is modulated by different types of ubiquitination and may lead to cell death, including apoptosis
Francisco Aguilar-Alonso et al.
Bioorganic & medicinal chemistry, 26(6), 1179-1188 (2017-12-17)
Linear ubiquitylation, in which ubiquitin units are covalently linked through N- and C-terminal amino acids, is a unique cellular signaling mechanism. This process is controlled by a single E3 ubiquitin ligase, the linear ubiquitin chain assembly complex (LUBAC), which is
Sebastian Kupka et al.
Cell reports, 16(9), 2271-2280 (2016-08-23)
Recruitment of the deubiquitinase CYLD to signaling complexes is mediated by its interaction with HOIP, the catalytically active component of the linear ubiquitin chain assembly complex (LUBAC). Here, we identify SPATA2 as a constitutive direct binding partner of HOIP that

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service