Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

MABS1293

Sigma-Aldrich

Anti-ING5 Antibody, clone 7A11

clone 7A11, from rat

Synonym(s):

Inhibitor of growth protein 5, p28ING5

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$444.00

$444.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
$444.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702

$444.00


Check Cart for Availability

Request a Bulk Order

biological source

rat

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

7A11, monoclonal

species reactivity

human

technique(s)

immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... ING5(84289)

General description

Inhibitor of growth protein 5 (UniProt Q8WYH8; also known as p28ING5) is encoded by the ING5 gene (Gene ID 84289) in human. ING5 is a member of the ING family of proteins known for their antiporliferation activity. ING proteins bind to methylated core histones through their PHD motif (a.a. 186-235 in ING5), and ING5 is known to target histone H3 on di- or trimethylated Lys4 (H3K4me2/3). ING5 associates with two histone acetylase (HAT) complexes, HBO1 and nucleosomal H3-specific MOZ/MORF, that interact with the MCM helicase and are essential for DNA replication during S phase. ING5 deregulations are reported in various cancers and ING5 located chromosome region is deleted in oral squamous cell carcinoma and odontogenic tumors. In addition, ING5 cytosolic localization is reported in some head and neck squamous cell carcinoma tumors and the cytosolic localization positively correlates with the progression of gastric cancer. ING5 also pormotes p53-dependent gene transcription by enhancing p53 acetylation in a p300-dependent manner. ING5 is shown to interact with p63 that mediates the self-renewal of epidermal stem cells, and ING5 knockdown causes differentiation of epidermal stem cells. Similar to other ING proteins, ING5 possesses a PHD (plant homeodomain) type zinc finger, an NCR (novel conserved region), a LZL (leucine zipper like) finger, and two NLS (nuclear localization sequence) motifs.

Specificity

Clone 7A11 detected shRNA-mediated ING5 downregulation in HEK293 cells (Linzen, U., et al. (2015). PLoS One. 10(4):e0123736).

Immunogen

GST-tagged recombinant human ING5 (Linzen, U., et al. (2015). PLoS One. 10(4):e0123736).

Application

Detect ING5 using this rat monoclonal Anti-ING5, clone 7A11 Antibody, Cat. No. MABS1293, validated for use in Immunocytochemistry, Immunoprecipitation, and Western Blotting.
Immunocytochemistry Analysis: A 1:250 dilution from a representative lot detected ING5 nuclear localization in HeLa cells.

Immunoprecipitation Analysis: A representative lot immunoprecipitated both Thr152-phosphorylated and non-Thr152-phosphorylated ING5, as well as exogenously expressed wild-type and T152A ING5 constructs (Linzen, U., et al. (2015). PLoS One. 10(4):e0123736).

Western Blotting Analysis: A representative lot detected ING5 in HCT116 and U2OS cell lysates, as well as downregulated ING5 in lysates from ING5 shRNAs-transfected HEK293 cells (Linzen, U., et al. (2015). PLoS One. 10(4):e0123736).
Research Category
Signaling

Quality

Evaluated by Western Blotting in Jurkat cell lysate.

Western Blotting Analysis: 1 µg/mL of this antibody detected ING5 in 10 µg of Jurkat cell lysate.

Target description

~31 kDa observed. 27.75 kDa (human ING5 isoform 1; UniProt Q8WYH8-1) calculated. The larger-than-calculated band size is consistent with that reported in the literature (Linzen, U., et al. (2015). PLoS One. 10(4):e0123736). Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified.
Purified rat IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service