MABS1257
Anti-APC4 Antibody, clone CIV1.1
clone CIV1.1, from mouse
Synonym(s):
Anaphase-promoting complex subunit 4, APC4, Cyclosome subunit 4
About This Item
biological source
mouse
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
CIV1.1, monoclonal
species reactivity
human
technique(s)
immunohistochemistry: suitable (paraffin)
isotype
IgG1κ
NCBI accession no.
UniProt accession no.
shipped in
ambient
target post-translational modification
unmodified
Gene Information
human ... ANAPC4(29945)
General description
Specificity
Immunogen
Application
Immunoprecipitation Analysis: A representative lot co-immunoprecipitated Cdc20, Mad2, and BubR1 with APC4 from prometaphase HeLa cells (Di Fiore, B., et al. (2015). Dev. Cell. 32(3):358-372).
Western Blotting Analysis: A representative lot detected APC4 in whole cell lysates, as well as in FLAG-tagged cyclin A and FLAG-tagged BubR1 immunoprecipitates from prometaphase HeLa cells (Di Fiore, B., et al. (2015). Dev. Cell. 32(3):358-372).
Quality
Immunohistochemistry Analysis: A 1:250 dilution of this antibody detected APC4 in human uterus tissue section.
Target description
Physical form
Other Notes
Not finding the right product?
Try our Product Selector Tool.
Storage Class
12 - Non Combustible Liquids
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service