Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABS1242

Sigma-Aldrich

Anti-CYP2J2 Antibody, clone 6-5-20-8

clone 6-5-20-8, from mouse

Synonym(s):

Cytochrome P450 2J2, Arachidonic acid epoxygenase, CYPIIJ2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$414.00

$414.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
$414.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$414.00


Check Cart for Availability

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

6-5-20-8, monoclonal

species reactivity

human, mouse

technique(s)

inhibition assay: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... CYP2J2(1573)

General description

Cytochrome P450 2J2 (UniProt: P51589; also known as EC:1.14.14.1, Arachidonic acid epoxygenase, CYPIIJ2) is encoded by the CYP2J2 gene (Gene ID: 1573) in human. Cytochrome P450 2J2 is an endoplasmic reticulum microsomal enzyme that metabolizes arachidonic acid predominantly via a NADPH-dependent olefin epoxidation to all four regioisomeric cis-epoxyeicosatrienoic acids. It is responsible for the epoxidation of endogenous cardiac arachidonic acid pools. It is highly expressed in heart and is present at lower levels in liver, ileum, jejunum, colon, and kidney. It is reported to activate the nuclear peroxisome proliferator-activated receptor alpha (PPAR alpha) and hence plays a role in lipid metabolism and inflammation. Overexpression of CYP2J2 in vivo is shown to reverse several pathological processes in animals, including hypertension, other cardiovascular pathologies, and insulin resistance.

Specificity

Clone 6-5-20-8 detects cytochrome p450 2J2 in human and murine species.

Immunogen

A full-length recombinant human cytochrome P450 2J2.

Application

Anti-CYP2J2, clone 6-5-20-8, Cat. No. MABS1242, is a mouse monoclonal antibody that targets cytochrome P450 2J2 and is tested for use in Inhibition Assay and Western Blotting.
Inhibition Analysis: A representative lot detected CYP2J2 in Inhibition applications (Xiao, Y.F., et. al. (2004). Mol Pharmacol. 66(6):1607-16).

Western Blotting Analysis: A representative lot detected CYP2J2 in Western Blotting applications (Xiao, Y.F., et. al. (2004). Mol Pharmacol. 66(6):1607-16).
Research Category
Signaling

Quality

Evaluated by Western Blotting in mouse heart tissue lysate.

Western Blotting Analysis: 1 µg/mL of this antibody detected CYP2J2 in 20 µg of mouse heart tissue lysate.

Target description

~52 kDa observed; 57.61 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG1 in PBS without azide.

Storage and Stability

Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service