Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

MABS1154

Sigma-Aldrich

Anti-Derlin-1 Antibody, clone 8G11.2

clone 8G11.2, from mouse

Synonym(s):

Degradation in endoplasmic reticulum protein 1, Der1-like protein 1, Derlin-1, DERtrin-1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$388.00

$388.00


Check Cart for Availability
A recombinant, preservative-free antibody is available for your target. Try ZRB1356

Request a Bulk Order

Select a Size

Change View
100 μG
$388.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$388.00


Check Cart for Availability
A recombinant, preservative-free antibody is available for your target. Try ZRB1356

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

8G11.2, monoclonal

species reactivity

human, mouse

technique(s)

immunocytochemistry: suitable
western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... DERL1(79139)

General description

Derlin-1 (UniProt Q9BUN8; also known as Degradation in endoplasmic reticulum protein 1, Der1-like protein 1, DERtrin-1) is encoded by the DERL1 (also known as DER1) gene (ORF UNQ243/PRO276; Gene ID 79139) in human. Derlins are integral ER membrane proteins that form homo-oligomers and heteromultimeric complexes with membrane-bound machinery, including the E3 ubiquitin ligase Hrd1 and its binding partner SEL1L, involved in ER-associated degradation (ERAD). Before ERAD substrates can be processed by the cytoplasmic ubiquitin-proteasome system, they need to be translocated across the ER membrane through an AAA ATPase p97/VCP-driven process. Derlins are inactive members of the rhomboid family of intramembrane proteases that mediate the recruitment of AAA ATPase p97/VCP through interaction via their C-terminal SHP boxes. Derlin-1 is proposed to contain four transmembrane segments (a.a. 23-43, 60-80, 106-126, 155-175), three cytoplasmic domains (a.a. 23-43, 60-80, 106-126, 155-175), and two lumenol (a.a. 44-59 & 127-154) domains.

Specificity

Expected to react with both spliced isoforms of human Derlin-1 reported by UniProt (Q9BUN8).

Immunogen

Epitope: C-terminal cytoplasmic domain.
GST-tagged recombinant human derlin-1 C-terminal cytoplasmic fragment.

Application

Immunocytochemistry Analysis: An 1:50 dilution from a representative lot detected Derlin-1 in HUVECs, A431 and HeLa cells.
Research Category
Signaling
Research Sub Category
Ubiquitin & Ubiquitin Metabolism
This Anti-Derlin-1 Antibody, clone 8G11.2 is validated for use in Western Blotting, Immunocytochemistry for the detection of Derlin-1.

Quality

Evaluated by Western Blotting in HEK293 cell lysate.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected Derlin-1 in 10 µg of HEK293 cell lysate.

Target description

~24 kDa observed. 28.80 kDa (isoform 1) and 26.42 kDa (isoform 2) calculated.

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG2bκ antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service