MABN759
Anti-DISC1 Antibody, clone 3D4
clone 3D4, 1 mg/mL, from mouse
Synonym(s):
Disrupted in schizophrenia 1 protein
Select a Size
About This Item
biological source
mouse
Quality Level
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
3D4, monoclonal
species reactivity
human
concentration
1 mg/mL
technique(s)
immunocytochemistry: suitable
immunohistochemistry: suitable (paraffin)
western blot: suitable
isotype
IgG1κ
NCBI accession no.
UniProt accession no.
shipped in
wet ice
target post-translational modification
unmodified
Gene Information
human ... DISC1(27185)
General description
Immunogen
Application
Western Blotting Analysis: A representative lot detected DISC1 in human NLF cell lysate (Leliveld, S. R., et al. (2008). Journal of Neuroscience. 28(15):3839-3845).
Immunocytochemistry Analysis: A representative lot detected DISC1 in transfected cells (Bader, V., et al. (2012). Human Molecular Genetics. 21(20):4406-4418).
Immunohistochemistry Analysis: A representative lot detected DISC1 in human cortical tissue sections (Seshadri, S. et al. (2010). PNAS. 107(12):5622-5627).
Immunohistochemistry Analysis: A representative lot detected DISC1 in human brain tissue (Bader, V., et al. (2012). Human Molecular Genetics. 21(20):4406-4418).
Neuroscience
Developmental Signaling
Quality
Immunohistochemistry Analysis: A 1:50 dilution of this antibody detected DISC1 in human cerebral cortex tissue.
Target description
Physical form
Storage and Stability
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class
12 - Non Combustible Liquids
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service