Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

MABN661

Sigma-Aldrich

Anti-QKI-5 Antibody, clone N195A/16

clone N195A/16, from mouse

Synonym(s):

Protein quaking, Hqk, HqkI

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

N195A/16, monoclonal

species reactivity

rat, human

species reactivity (predicted by homology)

mouse (based on 100% sequence homology)

technique(s)

immunohistochemistry: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... QKI(9444)

General description

QKI (Quaking) is an RNA-binding protein that plays a central role in myelinization. QKI acts by regulating pre-mRNA splicing, mRNA export, mRNA stability, and protein translation. It is regulated by alternative splicing. There are at least 6 known isoforms, and QKI-5 refers to isoforms 1, which is the ′canonical′ sequence, consisting of 341 amino acids.

Specificity

This antibody does not cross react with QKI-6, QKI-7, and QKI-7b. This antibody may cross react with other STAR proteins containing similar nuclear localization signals (Prof. J. Trimmer, Univerisity of California, Davis.).

Immunogen

Linear peptide corresponding to human QKI-5.

Application

Detect QKI using this mouse monoclonal antibody, Anti-QKI-5 Antibody, clone N195A/16 validated for use in IHC & western blotting.
Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected QKI-5 in human cerebellum and thalamus tissues, and in rat hippocampus tissue.

Western Blotting Analysis: A representative lot detected QKI-5 in COS cell lysates transiently transfected with Flag-tagged QKI-5 (Prof. J. Trimmer, University of California, Davis.).

Quality

Evaluated by Immunohistochemistry in rat cerebellum tissue.

Immunohistochemistry Analysis: A 1:50 dilution of this antibody detected QKI-5 in rat cerebellum tissue.

Target description

45 kDa calculated

Physical form

Format: Purified

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service