Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

MABN650

Sigma-Aldrich

Anti-Kirrel3 Antibody, short isoform 3, clone N320/48

clone N320/48, from mouse

Synonym(s):

Kin of IRRE-like protein 3, Kin of irregular chiasm-like protein 3, Processed kin of IRRE-like protein 3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

N320/48, monoclonal

species reactivity

human

technique(s)

immunohistochemistry: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... KIRREL3(84623)

General description

Kin of IRRE-like protein 3 (Kirrel3) or Kin of irregular chiasm-like protein 3 is expressed primarily in adult brain, bone marrow and stromal cells. Kirrel3 is thought to be involved in the hematopoietic supportive capacity of stroma cells, and the secreted extracellular domain of Kirrel3 is directly responsible for supporting hematopoietic stem cells.

Specificity

This antibody recognizes the cytoplasmic domain of Kirrel3, short isoform 3 and does not cross-react with other Kirrel3 isoforms.

Immunogen

Linear peptide corresponding to the cytoplasmic domain of mouse Kirrel3, short isoform 3.

Application

Detect Kin of IRRE-like protein 3 using this mouse monoclonal antibody, Anti-Kirrel3 Antibody, short isoform 3, clone N320/48 validated for use in western blotting & IHC.
Western Blotting Analysis: A representative lot detected Kirrel3, short isoform 3 in COS cell lysates transiently transfected with Kirrel3, short form, and demonstrated a loss of signal in COS cell lyasates transiently transfected with Kirrel3, long form or with untagged Kv2.1 plasmids (Prof. J. Trimmer, University of California, Davis.).
Immunohistochemistry Analysis: A 1:50-1,000 dilution from a representative lot detected Kirrel3, short isoform 3 in human thalamus and human bone marrow tissue.

Quality

Evaluated by Western Blotting in human brain tissue lysate.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected Kirrel3, short isoform 3 in 10 µg of human brain tissue lysate.

Target description

~ 75 kDa observed

Physical form

Format: Purified

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service