Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

MABN613

Sigma-Aldrich

Anti-Potassium channel Kv2.1 Antibody, clone L80/21

clone L80/21, from mouse

Synonym(s):

Potassium voltage-gated channel subfamily B member 1, Delayed rectifier potassium channel 1, DRK1, Voltage-gated potassium channel subunit Kv2.1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

antibody form

purified antibody

antibody product type

primary antibodies

clone

L80/21, monoclonal

species reactivity

rat, mouse

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG3κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

mouse ... Kcnb1(16500)

General description

Voltage-gated potassium channel subunit Kv2.1 (Kv2.1) is also called potassium voltage-gated channel subfamily B member 1 (KCNB1) or delayed rectifier potassium channel 1 (DRK1). Kv2.1 is a voltage-gated potassium ion channel that is expressed in the brain (cell bodies and dendrites of neurons but not axons, as well as Schwann cells), atria, ventricle, skeletal muscle, cochlea, retina, lung and pulmonary arteries. It is involved in maintaining membrane potential and modulating electrical excitability of both neurons and muscle cells. It is thought that Kv2.1 has a role in the muscle tone of the pulmonary system, as the expression Kv2.1 in pulmonary smooth muscle has been found to be reduced in chronic hypoxic pulmonary hypertension.

Specificity

This antibody recognizes the C-terminus of Potassium channel Kv2.1.

Immunogen

Epitope: C-terminus
Linear peptide corresponding to the C-terminus of rat Potassium channel Kv2.1.

Application

Detect Kv2.1 using this mouse monoclonal antibody, Anti-Potassium channel Kv2.1 Antibody, clone L80/21 validated for use in western blotting & IHC (Paraffin).
Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected Potassium channel Kv2.1 in rat frontal cortex tissue.

Western Blotting Analysis: A representative lot detected Potassium channel Kv2.1 in wild type mouse brain membrane tissue lysate, and demonstrated a loss of signal in Potassium channel Kv2.1 knock out mouse brain membrante tissue lysate (Prof. J. Trimmer, University of California, Davis.).
Research Category
Neuroscience
Research Sub Category
Synapse & Synaptic Biology

Quality

Evaluated by Western Blotting in rat brain tissue lysate.

Western Blotting Analysis: 1 µg/mL of this antibody detected Potassium channel Kv2.1 in 10 µg of rat brain tissue lysate.

Target description

~95 kDa observed

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG3κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Rat brain tissue lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service