MABN529
Anti-Neuronal acetylcholine receptor α-7, clone 6F12.2 Antibody
clone 6F12.2, from mouse
Synonym(s):
Neuronal acetylcholine receptor subunit alpha-7, acetylcholine receptor alpha-7, Neuronal acetylcholine receptor subunit alpha7, acetylcholine receptor alpha7, nAChRa7, nAChRa-7
About This Item
Recommended Products
biological source
mouse
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
6F12.2, monoclonal
species reactivity
mouse, human
technique(s)
immunohistochemistry: suitable
western blot: suitable
isotype
IgG1κ
NCBI accession no.
UniProt accession no.
shipped in
wet ice
target post-translational modification
unmodified
Gene Information
human ... CHRNA7(1139)
General description
Immunogen
Application
Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected Neuronal acetylcholine receptor α-7 in human cerebellum, cerebral cortex, and thalamic tissue.
Quality
Western Blotting Analysis: 2.0 µg/mL of this antibody detected Neuronal acetylcholine receptor α-7 in 10 µg of mouse cerebellum tissue lysate.
Target description
Physical form
Other Notes
Not finding the right product?
Try our Product Selector Tool.
Storage Class
12 - Non Combustible Liquids
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service