Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

MABN505

Sigma-Aldrich

Anti-Pink1 Antibody, clone N4/15

clone N4/15, from mouse

Synonym(s):

Serine/threonine-protein kinase PINK1, mitochondrial, BRPK, PTEN-induced putative kinase protein 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

N4/15, monoclonal

species reactivity

human, rat

technique(s)

western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... PINK1(65018)

General description

Mutations in the PTEN-induced kinase 1 (PINK1) gene located within the PARK6 locus on chromosome 1p35-p36 have recently been identified in patients with recessive early-onset Parkinson′s disease. Data suggests that PINK1 may be the second most common causative gene next to parkin in parkinsonism with the recessive mode of inheritance.

Specificity

This antibody recognizes the cytoplasmic domain of Pink1. Other Homologies: Mouse (81% Sequence Homology) and Rat (82% Sequence Homology)

Immunogen

Epitope: Cytoplasmic domain
Recombinant protein corresponding to the cytoplasmic domain of human Pink1.

Application

Anti-Pink1 Antibody, clone N4/15 is a highly specific mouse monoclonal antibody & that targets PINK1 & has been tested in western blotting.
Research Category
Neuroscience
Research Sub Category
Neurodegenerative Diseases
Western Blotting Analysis: 0.5 µg/mL from a representative lot detected Pink1 in 10 µg of human testis tissue lysate.

Immunohistochemistry Analysis: A representative lot detected Pink1 in rat caudate putamen tissue (Prof. J. Trimmer, University of California, Davis.).

Quality

Evaluated by Western Blotting in human skeletal muscle tissue lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected Pink1 in 10 µg of human skeletal muscle tissue lysate.

Target description

~60 kDa observed. Uniprot describes two isoforms at 63 kDa and 30 kDa The full length protein of Pink1 is observed at 63 kDa is the full length PINK1; however, it is proteolytically cleaved to a ~52 kDa form upon entry into the mitochondria (Jin, S. M., et al. (2010). J Cell Biol. 191(5):933-942.).

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Human skeletal muscle tissue lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Arash Aghajani Nargesi et al.
American journal of hypertension, 33(8), 765-774 (2020-03-18)
Mitochondria modulate endothelial cell (EC) function, but may be damaged during renal disease. We hypothesized that the ischemic and metabolic constituents of swine renovascular disease (RVD) induce mitochondrial damage and impair the function of renal artery ECs. Pigs were studied
Yi Zhang et al.
Journal of immunology (Baltimore, Md. : 1950), 192(11), 5296-5304 (2014-04-30)
High levels of inspired oxygen, hyperoxia, are frequently used in patients with acute respiratory failure. Hyperoxia can exacerbate acute respiratory failure, which has high mortality and no specific therapies. We identified novel roles for PTEN-induced putative kinase 1 (PINK1), a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service