Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABN487

Sigma-Aldrich

Anti-Cav3.2 Calcium Channel Subunit Antibody, clone N55/10

clone N55/10, from mouse

Synonym(s):

Cav3.2 Calcium Channel Subunit, Voltage-dependent T-type calcium channel subunit alpha-1H, Low-voltage-activated calcium channel alpha1 3.2 subunit, Voltage-gated calcium channel subunit alpha Cav3.2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

antibody form

purified antibody

antibody product type

primary antibodies

clone

N55/10, monoclonal

species reactivity

mouse

species reactivity (predicted by homology)

human (immunogen homology)

technique(s)

western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CACNA1H(8912)

General description

Voltage-gated calcium channel subunit alpha Cav3.2 is also called Voltage-dependent T-type calcium channel subunit alpha-1H and Low-voltage-activated calcium channel alpha 1 3.2 subunit. Voltage-gated calcium channel subunit alpha Cav3.2 and other voltage-sensitive calcium channels (VSCC) are involved in a variety of calcium-dependent processes, including muscle contraction, hormone or neurotransmitter release, gene expression, cell motility, cell division and cell death. Voltage-gated calcium channel subunit alpha Cav3.2 is expressed in kidney, liver, heart, brain, and isoform 2 appears to be testis specific. Voltage-gated calcium channel subunit alpha Cav3.2 defects can increase susceptibility to epilepsy, idiopathic generalized type 6 (EIG6) and epilepsy, childhood absence type 6 (ECA6).

Specificity

This antibody does not cross react with human Cav3.1 Calcium Channel Subunit (Prof. J. Trimmer, University of California, Davis.).

Immunogen

Recombinant protein corresponding to human Cav3.2 Calcium Channel Subunit.

Application

Detect Cav3.2 using this mouse monoclonal antibody, Anti-Cav3.2 Calcium Channel Subunit Antibody & clone N55/10 validated for use in western blotting.
Immunohistochemistry Analysis: A representative lot detected Cav3.2 Calcium Channel Subunit in wild type mice and demonstrated a loss of signal in Cav3.2 knock out mice (Prof. J. Trimmer, University of California, Davis.).

Western Blotting Analysis: A representative lot detected Cav3.2 Calcium Channel Subunit in HEK293 cell lysates expressing Flag-tagged Cav3.2 and demonstrated a loss of signal in HEK293 cell lysates expressing Flag-tagged Kv2.1 and Flag-tagged Cav3.1 (Prof. J. Trimmer, University of California, Davis.).
Research Category
Neuroscience
Research Sub Category
Synapse & Synaptic Biology

Quality

Evaluated by Western Blotting in NIH/3T3 cell lysate.

Western Blotting Analysis: 1 µg/mL of this antibody detected Cav3.2 Calcium Channel Subunit in 10 µg of NIH/3T3 cell lysate.

Target description

~260 kDa observed

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
NIH/3T3 cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service