Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

MABN478

Sigma-Aldrich

Anti-Spred-1 Antibody, clone M23-P2G3

clone M23-P2G3, from mouse, purified by affinity chromatography

Synonym(s):

Sprouty-related, EVH1 domain-containing protein 1, Spred-1, hSpred1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$444.00

$444.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
$444.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$444.00


Check Cart for Availability

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

M23-P2G3, monoclonal

purified by

affinity chromatography

species reactivity

rat, mouse

species reactivity (predicted by homology)

human (based on 100% sequence homology)

technique(s)

immunocytochemistry: suitable
western blot: suitable

isotype

IgG1λ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... SPRED1(161742)

General description

Sprouty-related, EVH1 domain-containing protein 1 (SPRED1) is a tyrosine kinase substrate which has a cysteine-rich domain which resembles the SPR domain at the carboxy terminus. SPRED1 associated with Ras blocks phosphorylation of Raf and disrupts MAP kinase signaling during the differentiation of neuronal cells and myocytes. A mutation in SPRED1 has been linked to Lugius syndrome, an autosomal dominant RAS-MAPK pathway syndrome.

Immunogen

Linear peptide corresponding to human Spred-1.

Application

Detect Spred-1 using this mouse monoclonal antibody, Anti-Spred-1 Antibody, clone M23-P2G3 validated for use in western blotting & ICC.
Immunocytochemistry Analysis: A representative lot detected Spred-1 in HeLa cells.
Research Category
Neuroscience
Research Sub Category
Developmental Signaling

Quality

Evaluated by Western Blotting in mouse brain tissue lysate.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected Spred-1 in 10 µg of mouse brain tissue lysate.

Target description

~ 60 kDa observed

Physical form

Protein G Purified
Purified mouse monoclonal IgG1λ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service