Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

MABN462

Sigma-Aldrich

Anti-ASIC1 Antibody, clone N271/44

clone N271/44, from mouse

Synonym(s):

Acid-sensing ion channel 1, ASIC1, Acid-sensing ion channel, Amiloride-sensitive cation channel 2, neuronal, Brain sodium channel 2, BNaC2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

N271/44, monoclonal

species reactivity

rat, mouse

technique(s)

immunohistochemistry: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

General description

Acid-sensing ion channels (ASICs) are broadly distributed in the nervous system where they contribute to certain sensory processes. In particular, they survey moderate decreases in extracellular pH as a means to regulate acid levels and acid-induced pain. ASICs thereby serve as a popular target for the treatment of acidosis. The acid-sensing ion channel 1 consists of a trimeric protein, where each subunit is composed of two transmembrane spanning helices, a highly organized ectodomain, and an intracellular N-terminus and C-terminus.

Specificity

This antibody recognizes the C-terminus of ASIC1.

Immunogen

Epitope: C-terminus
Recombinant protein corresponding to the C-terminus of mouse ASIC1.

Application

Anti-ASIC1 Antibody, clone N271/44 detects level of ASIC1 & has been published & validated for use in Western Blotting & IHC.
Research Category
Neuroscience
Research Sub Category
Developmental Neuroscience
Western Blot: A representative lot detected ASIC1 in WT mice, but demonstrated a loss of signal in ASIC1 knockout mice.

Immunohistochemistry Analysis: A representative lot detected ASIC1 in rat brain tissue (Prof. J. Trimmer, University of California, Davis).

Quality

Evaluated by Western Blot in rat brain tissue lysate.

Western Blot Analysis: 1 µg/mL of this antibody detected ASIC1 in 10 µg of rat brain tissue lysate.

Target description

~60 kDa observed. Two isoforms at ~60 kDa and ~62 kDa are known to exist.

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Rat brain tissue lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hongfei Ge et al.
Research (Washington, D.C.), 6, 0105-0105 (2023-06-05)
Cell replacement therapy using neural progenitor cells (NPCs) has been shown to be an effective treatment for ischemic stroke. However, the therapeutic effect is unsatisfactory due to the imbalanced homeostasis of the local microenvironment after ischemia. Microenvironmental acidosis is a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service