Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

MABN461

Sigma-Aldrich

Anti-Dicer1 Antibody, clone N167/7

clone N167/7, from mouse

Synonym(s):

Endoribonuclease Dicer, Double-strand-specific ribonuclease mDCR-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

antibody form

purified antibody

antibody product type

primary antibodies

clone

N167/7, monoclonal

species reactivity

rat, human, mouse

technique(s)

immunohistochemistry: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

mouse ... Dicer1(192119)

General description

RNA interference (RNAi) is mechanism for post-transcriptional gene silencing that leads to the loss of the activity of the gene. RNAi is activated when long dsRNAs are cleaved by the enzyme Dicer1 (RNase III family of dsRNA-specific endonucleases) into 21-23 nucleotide siRNA (small interfering RNA). These siRNAs bind proteins to form the RISC (RNA-induced silencing complex) which is activated when the double stranded siRNAs are unzipped to single stranded siRNAs. Activated RISC recognizes and binds complementary sequences in mRNA. The mRNA is cleaved and the overall effect is an inhibition of the protein expression and a loss of the gene activity. Dicer1 along with the Argonaute family of proteins (Ago1 and Ago2) share the same domain PIWI and PAZ domain that provide endonuclease activity to RNA-induced silencing complexes (RISC).

Immunogen

Recombinant protein corresponding to mouse Dicer1.

Application

Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected Dicer1 in human ovarian adenocarcinoma tissues and in rat cerebellum tissue.

Western Blot Analysis: A representative lot from an independent laboratory detected Dicer1 in adult rat brain membrane and in WT mouse brain lysate, and demonstrated a loss of signal in Dicer1 knockout mouse brain lysate.
Research Category
Neuroscience
Research Sub Category
Developmental Neuroscience
This Dicer1 antibody is validated for use in WB & IHC for the detection of the Dicer1 protein.

Quality

Evaluated by Western Blotting in MCF-7 cell lysate.

Western Blotting Analysis: 2 µg/mL of this antibody detected Dicer1 in 10 µg of MCF-7 cell lysate.

Target description

~217 kDa observed

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
MCF-7 cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Neeraj K Aryal et al.
Proceedings of the National Academy of Sciences of the United States of America, 116(3), 960-969 (2018-12-30)
DICER1 gene alterations and decreased expression are associated with developmental disorders and diseases in humans. Oscillation of Dicer1 phosphorylation and dephosphorylation regulates its function during the oocyte-to-embryo transition in Caenorhabditis elegans Dicer1 is also phosphorylated upon FGF stimulation at conserved

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service