Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

MABN452

Sigma-Aldrich

Anti-Adenosine Deaminase Antibody, clone 9H12.1

clone 9H12.1, from mouse

Synonym(s):

Adenosine deaminase CECR1, Cat eye syndrome critical region protein 1, CECR1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

9H12.1, monoclonal

species reactivity

human

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgMκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

General description

Adenosine deaminase, also known as ADA, or Cat eye syndrome critical region protein 1, and encoded by the gene CECR1/ADA2/ADGF/IDGFL, is an extracellular enzyme that works on adenosine and causes the degradation of adenosine to inosine and free NH3. Adenosine deaminase is an extracellular secreted enzyme that binds to cell surfaces via proteoglycans. The binding appears to mediate cellular proliferation and differentiation in many systems, though this activity is independent of its deaminase activity. Adenosine deaminase is highly present in blood plasma and widely expressed in adult and fetal tissues. While there many clinical deficiencies associated with adenosine deaminase (ADA) mutations, the level of ADA activity in the blood and fluids is used as a biomarker to help distinguish various diseases. For instance, the level of activity in lung fluid is correlated with tuberculosis infection and used to distinguish it from malignant pleural effusion (MPE). The ADA level in CSF in patients with meningitis also helps to identify tuberculosis as the cause as opposed to another agent or condition.

Immunogen

GST-tagged recombinant protein corresponding to human Adenosine Deaminase.

Application

Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected Adenosine Deaminase in human placenta tissue.
Research Category
Neuroscience
Research Sub Category
Developmental Signaling
This Anti-Adenosine Deaminase Antibody, clone 9H12.1 is validated for use in western blotting & IHC (Paraffin) for the detection of Adenosine Deaminase.

Quality

Evaluated by Western Blotting in human placenta tissue lysate.

Western Blotting Analysis: A 1:500 dilution of this antibody detected Adenosine Deaminase in 10 µg of human placenta tissue lysate.

Target description

~ 60 kDa observed

Physical form

Format: Purified
Purified mouse monoclonal IgMκ in buffer containing PBS with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service